Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50778604-50778877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK3
Entrez Gene ID
1795
Gene Name
dedicator of cytokinesis 3
Gene Aliases
MOCA, PBP
UniGene
Hs.476284
Gene Chromosome Location
Chr.3: 50674969-51384198 on Build GRCh38
Transcripts
NM_004947.4 XM_005264914.3 XM_005264915.3 XM_005264916.3 XM_005264917.3 XM_005264918.3 XM_006713008.3 XM_006713009.3 XM_006713010.3 XM_017005825.1 XM_017005826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5918061 c.64C>T p.P22S Chr.3: 50778701-50778701 on Build GRCh38
460762 c.67C>G p.Q23E Chr.3: 50778704-50778704 on Build GRCh38
4678998 c.99_100delCC p.Q34fs*3 Chr.3: 50778736-50778737 on Build GRCh38
5715725 c.56G>A p.G19E Chr.3: 50778693-50778693 on Build GRCh38
5723673 c.52C>T p.R18* Chr.3: 50778689-50778689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777299900 Chr.3: 50778794-50778794 on Build GRCh38
rs975836407 Chr.3: 50778839-50778839 on Build GRCh38
rs771650789 Chr.3: 50778774-50778774 on Build GRCh38
rs778551371 Chr.3: 50778758-50778758 on Build GRCh38
rs1042547879 Chr.3: 50778757-50778757 on Build GRCh38
rs767459845 Chr.3: 50778679-50778679 on Build GRCh38
rs540330072 Chr.3: 50778685-50778685 on Build GRCh38
rs777400073 Chr.3: 50778695-50778695 on Build GRCh38
rs200645618 Chr.3: 50778698-50778698 on Build GRCh38
rs745374676 Chr.3: 50778773-50778773 on Build GRCh38
rs13067267 Chr.3: 50778631-50778631 on Build GRCh38
rs572280976 Chr.3: 50778821-50778821 on Build GRCh38
rs553918838 Chr.3: 50778650-50778650 on Build GRCh38
rs912756031 Chr.3: 50778706-50778706 on Build GRCh38
rs553447000 Chr.3: 50778830-50778830 on Build GRCh38
rs756847907 Chr.3: 50778739-50778739 on Build GRCh38
rs767290600 Chr.3: 50778678-50778678 on Build GRCh38
rs184886480 Chr.3: 50778646-50778646 on Build GRCh38
rs944209035 Chr.3: 50778785-50778785 on Build GRCh38
rs200405924 Chr.3: 50778690-50778690 on Build GRCh38

View Full Product Details