Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44856968-44857239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFTUD2
Entrez Gene ID
9343
Gene Name
elongation factor Tu GTP binding domain containing 2
Gene Aliases
MFDGA, MFDM, SNRNP116, Snrp116, Snu114, U5-116KD
UniGene
Hs.151787
Gene Chromosome Location
Chr.17: 44850287-44899625 on Build GRCh38
Transcripts
NM_001142605.1 NM_001258353.1 NM_001258354.1 NM_004247.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6277895 c.2001G>A p.V667V Chr.17: 44857119-44857119 on Build GRCh38
3518147 c.2013C>T p.S671S Chr.17: 44857107-44857107 on Build GRCh38
5946733 c.1971C>T p.D657D Chr.17: 44857149-44857149 on Build GRCh38
5594529 c.2012C>T p.S671F Chr.17: 44857108-44857108 on Build GRCh38
6241258 c.1994C>T p.T665M Chr.17: 44857126-44857126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953652456 Chr.17: 44856998-44856998 on Build GRCh38
rs772825325 Chr.17: 44857155-44857155 on Build GRCh38
rs58089352 Chr.17: 44857047-44857047 on Build GRCh38
rs765835040 Chr.17: 44857158-44857158 on Build GRCh38
rs749893961 Chr.17: 44857051-44857051 on Build GRCh38
rs551609901 Chr.17: 44857021-44857022 on Build GRCh38
rs759131015 Chr.17: 44857184-44857184 on Build GRCh38
rs775468874 Chr.17: 44857026-44857026 on Build GRCh38
rs372893019 Chr.17: 44857052-44857052 on Build GRCh38
rs141842808 Chr.17: 44857086-44857086 on Build GRCh38
rs764579662 Chr.17: 44857186-44857186 on Build GRCh38
rs968732516 Chr.17: 44857037-44857037 on Build GRCh38
rs768525340 Chr.17: 44857045-44857045 on Build GRCh38
rs6503405 Chr.17: 44857004-44857004 on Build GRCh38
rs781402078 Chr.17: 44857087-44857087 on Build GRCh38
rs776271665 Chr.17: 44857165-44857165 on Build GRCh38
rs753439433 Chr.17: 44857060-44857060 on Build GRCh38
rs774001446 Chr.17: 44857138-44857138 on Build GRCh38
rs780448978 Chr.17: 44857119-44857119 on Build GRCh38
rs752540647 Chr.17: 44857201-44857201 on Build GRCh38
rs863224868 Chr.17: 44857073-44857073 on Build GRCh38
rs767110966 Chr.17: 44857048-44857048 on Build GRCh38
rs752121790 Chr.17: 44857080-44857080 on Build GRCh38
rs770193739 Chr.17: 44857113-44857113 on Build GRCh38
rs1046396361 Chr.17: 44857142-44857142 on Build GRCh38
rs144798973 Chr.17: 44857078-44857078 on Build GRCh38
rs965660832 Chr.17: 44857034-44857034 on Build GRCh38
rs767644587 Chr.17: 44857026-44857027 on Build GRCh38
rs368215462 Chr.17: 44857125-44857125 on Build GRCh38
rs771872448 Chr.17: 44857146-44857146 on Build GRCh38
rs374850141 Chr.17: 44857061-44857061 on Build GRCh38
rs201911879 Chr.17: 44857172-44857172 on Build GRCh38
rs921168119 Chr.17: 44857160-44857160 on Build GRCh38
rs770138545 Chr.17: 44857025-44857025 on Build GRCh38
rs185635465 Chr.17: 44857187-44857187 on Build GRCh38
rs147121628 Chr.17: 44857137-44857137 on Build GRCh38
rs1013016723 Chr.17: 44857007-44857007 on Build GRCh38
rs765991243 Chr.17: 44857058-44857058 on Build GRCh38
rs575007031 Chr.17: 44856999-44856999 on Build GRCh38
rs745906919 Chr.17: 44857104-44857104 on Build GRCh38
rs894661242 Chr.17: 44857006-44857006 on Build GRCh38
rs774346019 Chr.17: 44857046-44857046 on Build GRCh38
rs760199934 Chr.17: 44857157-44857157 on Build GRCh38
rs757826608 Chr.17: 44857198-44857198 on Build GRCh38
rs762824210 Chr.17: 44857028-44857028 on Build GRCh38

View Full Product Details