Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28524515-28524788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN1
Entrez Gene ID
8456
Gene Name
forkhead box N1
Gene Aliases
FKHL20, RONU, WHN
UniGene
Hs.58611
Gene Chromosome Location
Chr.17: 28506211-28538900 on Build GRCh38
Transcripts
NM_003593.2 XM_005258046.3 XM_011525358.1 XM_011525359.1 XM_011525362.1 XM_011525368.2 XM_017025228.1 XM_017025229.1 XM_017025230.1 XM_017025231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3771297 c.213G>A p.A71A Chr.17: 28524592-28524592 on Build GRCh38
4638978 c.360C>T p.H120H Chr.17: 28524739-28524739 on Build GRCh38
1128782 c.207C>T p.R69R Chr.17: 28524586-28524586 on Build GRCh38
6538684 c.178A>G p.T60A Chr.17: 28524557-28524557 on Build GRCh38
976882 c.234C>A p.V78V Chr.17: 28524613-28524613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026597581 Chr.17: 28524720-28524720 on Build GRCh38
rs142248984 Chr.17: 28524605-28524605 on Build GRCh38
rs373442344 Chr.17: 28524739-28524739 on Build GRCh38
rs373326758 Chr.17: 28524591-28524591 on Build GRCh38
rs868443961 Chr.17: 28524633-28524633 on Build GRCh38
rs200786966 Chr.17: 28524541-28524541 on Build GRCh38
rs751230017 Chr.17: 28524745-28524745 on Build GRCh38
rs745766812 Chr.17: 28524675-28524675 on Build GRCh38
rs777608417 Chr.17: 28524573-28524573 on Build GRCh38
rs567680104 Chr.17: 28524645-28524645 on Build GRCh38
rs938556199 Chr.17: 28524560-28524560 on Build GRCh38
rs753353730 Chr.17: 28524545-28524545 on Build GRCh38
rs769969790 Chr.17: 28524585-28524585 on Build GRCh38
rs138295148 Chr.17: 28524700-28524700 on Build GRCh38
rs754433537 Chr.17: 28524742-28524742 on Build GRCh38
rs1015604026 Chr.17: 28524691-28524691 on Build GRCh38
rs756535565 Chr.17: 28524756-28524757 on Build GRCh38
rs777223855 Chr.17: 28524639-28524639 on Build GRCh38
rs765915993 Chr.17: 28524638-28524638 on Build GRCh38
rs557541901 Chr.17: 28524741-28524741 on Build GRCh38
rs757771641 Chr.17: 28524542-28524542 on Build GRCh38
rs746851611 Chr.17: 28524650-28524650 on Build GRCh38
rs761166991 Chr.17: 28524714-28524714 on Build GRCh38
rs781323725 Chr.17: 28524662-28524662 on Build GRCh38
rs544854723 Chr.17: 28524592-28524592 on Build GRCh38
rs951831368 Chr.17: 28524649-28524649 on Build GRCh38
rs368024968 Chr.17: 28524538-28524538 on Build GRCh38
rs746723858 Chr.17: 28524577-28524577 on Build GRCh38
rs747815915 Chr.17: 28524558-28524558 on Build GRCh38
rs770996675 Chr.17: 28524580-28524580 on Build GRCh38
rs200888774 Chr.17: 28524539-28524539 on Build GRCh38
rs772403336 Chr.17: 28524711-28524711 on Build GRCh38
rs757155505 Chr.17: 28524752-28524752 on Build GRCh38
rs142380525 Chr.17: 28524550-28524550 on Build GRCh38
rs144301161 Chr.17: 28524761-28524761 on Build GRCh38
rs762103229 Chr.17: 28524601-28524601 on Build GRCh38
rs182334974 Chr.17: 28524740-28524740 on Build GRCh38
rs764774942 Chr.17: 28524631-28524631 on Build GRCh38
rs775611762 Chr.17: 28524587-28524587 on Build GRCh38
rs149614149 Chr.17: 28524754-28524754 on Build GRCh38
rs1013013858 Chr.17: 28524607-28524607 on Build GRCh38
rs552574496 Chr.17: 28524562-28524562 on Build GRCh38
rs756136088 Chr.17: 28524757-28524757 on Build GRCh38
rs776666522 Chr.17: 28524582-28524582 on Build GRCh38
rs1019777033 Chr.17: 28524749-28524749 on Build GRCh38
rs774400145 Chr.17: 28524701-28524701 on Build GRCh38
rs2071587 Chr.17: 28524584-28524584 on Build GRCh38
rs374082998 Chr.17: 28524658-28524658 on Build GRCh38
rs750882115 Chr.17: 28524611-28524611 on Build GRCh38
rs150894024 Chr.17: 28524615-28524615 on Build GRCh38
rs770062805 Chr.17: 28524678-28524678 on Build GRCh38
rs749272531 Chr.17: 28524698-28524698 on Build GRCh38
rs138510671 Chr.17: 28524604-28524604 on Build GRCh38
rs773751011 Chr.17: 28524713-28524713 on Build GRCh38
rs1041633589 Chr.17: 28524544-28524544 on Build GRCh38
rs372060062 Chr.17: 28524704-28524704 on Build GRCh38
rs199685259 Chr.17: 28524553-28524553 on Build GRCh38
rs775629575 Chr.17: 28524679-28524679 on Build GRCh38
rs752348608 Chr.17: 28524632-28524632 on Build GRCh38
rs781127331 Chr.17: 28524753-28524753 on Build GRCh38

View Full Product Details