Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27658023-27658282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDNF
Entrez Gene ID
627
Gene Name
brain derived neurotrophic factor
Gene Aliases
ANON2, BULN2
UniGene
Hs.502182
Gene Chromosome Location
Chr.11: 27654893-27722058 on Build GRCh38
Transcripts
NM_001143805.1 NM_001143806.1 NM_001143807.1 NM_001143808.1 NM_001143809.1 NM_001143810.1 NM_001143811.1 NM_001143812.1 NM_001143813.1 NM_001143814.1 NM_001143816.1 NM_001709.4 NM_170731.4 NM_170732.4 NM_170733.3 NM_170734.3 NM_170735.5 XM_011520280.2 NR_002832.2 NR_033312.1 NR_033313.1 NR_033314.1 NR_033315.1

Gene Symbol
BDNF-AS
Entrez Gene ID
497258
Gene Name
BDNF antisense RNA
Gene Aliases
ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049
UniGene
Hs.675323
Gene Chromosome Location
Chr.11: 27506852-27698171 on Build GRCh38
Transcripts
NM_001143805.1 NM_001143806.1 NM_001143807.1 NM_001143808.1 NM_001143809.1 NM_001143810.1 NM_001143811.1 NM_001143812.1 NM_001143813.1 NM_001143814.1 NM_001143816.1 NM_001709.4 NM_170731.4 NM_170732.4 NM_170733.3 NM_170734.3 NM_170735.5 XM_011520280.2 NR_002832.2 NR_033312.1 NR_033313.1 NR_033314.1 NR_033315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1353340 c.404G>A p.R135Q Chr.11: 27658185-27658185 on Build GRCh38
5731926 c.748G>A p.E250K Chr.11: 27658063-27658063 on Build GRCh38
403634 c.512C>T p.T171M Chr.11: 27658077-27658077 on Build GRCh38
1246067 c.730G>A p.G244R Chr.11: 27658081-27658081 on Build GRCh38
4865180 c.524C>T p.S175L Chr.11: 27658086-27658086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750743108 Chr.11: 27658202-27658202 on Build GRCh38
rs755151584 Chr.11: 27658242-27658242 on Build GRCh38
rs148736382 Chr.11: 27658244-27658244 on Build GRCh38
rs754270781 Chr.11: 27658077-27658077 on Build GRCh38
rs777804423 Chr.11: 27658109-27658109 on Build GRCh38
rs763510956 Chr.11: 27658190-27658190 on Build GRCh38
rs1048220 Chr.11: 27658191-27658191 on Build GRCh38
rs766669848 Chr.11: 27658226-27658226 on Build GRCh38
rs751698045 Chr.11: 27658234-27658234 on Build GRCh38
rs760274076 Chr.11: 27658148-27658148 on Build GRCh38
rs758638310 Chr.11: 27658213-27658213 on Build GRCh38
rs764711746 Chr.11: 27658076-27658076 on Build GRCh38
rs2353512 Chr.11: 27658115-27658115 on Build GRCh38
rs17852698 Chr.11: 27658211-27658211 on Build GRCh38
rs540912088 Chr.11: 27658153-27658153 on Build GRCh38
rs935219250 Chr.11: 27658256-27658256 on Build GRCh38
rs774985498 Chr.11: 27658147-27658147 on Build GRCh38
rs1048221 Chr.11: 27658185-27658185 on Build GRCh38
rs761108714 Chr.11: 27658070-27658070 on Build GRCh38
rs376008850 Chr.11: 27658082-27658082 on Build GRCh38
rs77787410 Chr.11: 27658057-27658057 on Build GRCh38
rs747058681 Chr.11: 27658100-27658100 on Build GRCh38
rs750030464 Chr.11: 27658086-27658086 on Build GRCh38
rs1049779568 Chr.11: 27658173-27658173 on Build GRCh38
rs771802172 Chr.11: 27658142-27658142 on Build GRCh38
rs201468555 Chr.11: 27658102-27658102 on Build GRCh38
rs765594245 Chr.11: 27658200-27658200 on Build GRCh38
rs144235869 Chr.11: 27658118-27658118 on Build GRCh38
rs749232626 Chr.11: 27658112-27658112 on Build GRCh38
rs766339291 Chr.11: 27658207-27658207 on Build GRCh38
rs774229943 Chr.11: 27658116-27658116 on Build GRCh38
rs201119590 Chr.11: 27658195-27658195 on Build GRCh38
rs370102323 Chr.11: 27658085-27658085 on Build GRCh38
rs990473824 Chr.11: 27658114-27658114 on Build GRCh38

View Full Product Details