Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58622672-58622943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTF
Entrez Gene ID
1270
Gene Name
ciliary neurotrophic factor
Gene Aliases
HCNTF
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58622673-58625733 on Build GRCh38
Transcripts
NM_000614.3 NR_024091.1

Gene Symbol
ZFP91-CNTF
Entrez Gene ID
386607
Gene Name
ZFP91-CNTF readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58579114-58625733 on Build GRCh38
Transcripts
NM_000614.3 NR_024091.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274594 c.82C>T p.R28C Chr.11: 58622834-58622834 on Build GRCh38
360761 c.97G>A p.A33T Chr.11: 58622849-58622849 on Build GRCh38
5050920 c.57C>A p.R19R Chr.11: 58622809-58622809 on Build GRCh38
3368440 c.83G>A p.R28H Chr.11: 58622835-58622835 on Build GRCh38
928897 c.39T>C p.R13R Chr.11: 58622791-58622791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768790834 Chr.11: 58622823-58622823 on Build GRCh38
rs752850472 Chr.11: 58622698-58622698 on Build GRCh38
rs375396915 Chr.11: 58622741-58622741 on Build GRCh38
rs888678462 Chr.11: 58622817-58622817 on Build GRCh38
rs755622885 Chr.11: 58622867-58622867 on Build GRCh38
rs78916864 Chr.11: 58622890-58622890 on Build GRCh38
rs779488833 Chr.11: 58622734-58622734 on Build GRCh38
rs143821189 Chr.11: 58622857-58622857 on Build GRCh38
rs751150725 Chr.11: 58622849-58622849 on Build GRCh38
rs772318962 Chr.11: 58622756-58622756 on Build GRCh38
rs772636468 Chr.11: 58622815-58622815 on Build GRCh38
rs369596489 Chr.11: 58622731-58622731 on Build GRCh38
rs759631204 Chr.11: 58622753-58622753 on Build GRCh38
rs149242903 Chr.11: 58622808-58622808 on Build GRCh38
rs34131586 Chr.11: 58622865-58622865 on Build GRCh38
rs746423555 Chr.11: 58622878-58622878 on Build GRCh38
rs747249412 Chr.11: 58622822-58622822 on Build GRCh38
rs201872988 Chr.11: 58622782-58622782 on Build GRCh38
rs377441103 Chr.11: 58622720-58622720 on Build GRCh38
rs538707475 Chr.11: 58622810-58622810 on Build GRCh38
rs201519587 Chr.11: 58622791-58622791 on Build GRCh38
rs1041143948 Chr.11: 58622792-58622792 on Build GRCh38
rs758777968 Chr.11: 58622886-58622886 on Build GRCh38
rs751279439 Chr.11: 58622799-58622799 on Build GRCh38
rs766216220 Chr.11: 58622844-58622844 on Build GRCh38
rs746500294 Chr.11: 58622737-58622737 on Build GRCh38
rs779648726 Chr.11: 58622798-58622798 on Build GRCh38
rs139489162 Chr.11: 58622778-58622778 on Build GRCh38
rs761817013 Chr.11: 58622790-58622790 on Build GRCh38
rs764096705 Chr.11: 58622709-58622709 on Build GRCh38
rs977917974 Chr.11: 58622883-58622883 on Build GRCh38
rs776884845 Chr.11: 58622832-58622832 on Build GRCh38
rs371899433 Chr.11: 58622914-58622914 on Build GRCh38
rs758995566 Chr.11: 58622914-58622915 on Build GRCh38
rs760889934 Chr.11: 58622701-58622701 on Build GRCh38
rs75769623 Chr.11: 58622820-58622820 on Build GRCh38
rs779473520 Chr.11: 58622874-58622874 on Build GRCh38
rs201478772 Chr.11: 58622836-58622836 on Build GRCh38
rs779340530 Chr.11: 58622813-58622813 on Build GRCh38
rs755779702 Chr.11: 58622745-58622746 on Build GRCh38
rs529701662 Chr.11: 58622793-58622793 on Build GRCh38
rs758175765 Chr.11: 58622794-58622794 on Build GRCh38
rs200245057 Chr.11: 58622776-58622776 on Build GRCh38
rs778784352 Chr.11: 58622728-58622728 on Build GRCh38
rs777362153 Chr.11: 58622789-58622789 on Build GRCh38
rs368279696 Chr.11: 58622856-58622856 on Build GRCh38
rs749979700 Chr.11: 58622730-58622730 on Build GRCh38
rs761605079 Chr.11: 58622834-58622834 on Build GRCh38
rs779444968 Chr.11: 58622801-58622802 on Build GRCh38
rs769023999 Chr.11: 58622911-58622911 on Build GRCh38
rs753358998 Chr.11: 58622805-58622807 on Build GRCh38
rs377547716 Chr.11: 58622750-58622750 on Build GRCh38
rs565152907 Chr.11: 58622775-58622775 on Build GRCh38
rs370237695 Chr.11: 58622740-58622740 on Build GRCh38
rs747196357 Chr.11: 58622891-58622891 on Build GRCh38
rs139149169 Chr.11: 58622835-58622835 on Build GRCh38
rs756990672 Chr.11: 58622725-58622725 on Build GRCh38
rs754467848 Chr.11: 58622807-58622807 on Build GRCh38

View Full Product Details