Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200818694-200818953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BZW1
Entrez Gene ID
9689
Gene Name
basic leucine zipper and W2 domains 1
Gene Aliases
BZAP45, Nbla10236
UniGene
Hs.355983
Gene Chromosome Location
Chr.2: 200811546-200823851 on Build GRCh38
Transcripts
NM_001207067.1 NM_001207069.1 NM_001321688.1 NM_001321690.1 NM_001321691.1 NM_001321693.1 NM_001321694.1 NM_014670.3 XM_017005393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1614128 c.845T>G p.M282R Chr.2: 200818780-200818780 on Build GRCh38
1404422 c.943delA p.N317fs*11 Chr.2: 200818782-200818782 on Build GRCh38
5069884 c.846_847insA p.N285fs*18 Chr.2: 200818781-200818782 on Build GRCh38
1591664 c.1053G>A p.K351K Chr.2: 200818892-200818892 on Build GRCh38
5128201 c.846_847insAA p.N285fs*7 Chr.2: 200818781-200818782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771335930 Chr.2: 200818775-200818775 on Build GRCh38
rs779726236 Chr.2: 200818888-200818888 on Build GRCh38
rs374451772 Chr.2: 200818773-200818773 on Build GRCh38
rs778849072 Chr.2: 200818761-200818761 on Build GRCh38
rs776829596 Chr.2: 200818803-200818803 on Build GRCh38
rs747924141 Chr.2: 200818763-200818763 on Build GRCh38
rs746064938 Chr.2: 200818788-200818790 on Build GRCh38
rs749331552 Chr.2: 200818725-200818729 on Build GRCh38
rs892189564 Chr.2: 200818904-200818904 on Build GRCh38
rs775957419 Chr.2: 200818865-200818865 on Build GRCh38
rs755744469 Chr.2: 200818871-200818871 on Build GRCh38
rs376962712 Chr.2: 200818835-200818835 on Build GRCh38
rs567838118 Chr.2: 200818924-200818924 on Build GRCh38
rs766917456 Chr.2: 200818923-200818923 on Build GRCh38
rs765790920 Chr.2: 200818820-200818820 on Build GRCh38
rs370998325 Chr.2: 200818753-200818753 on Build GRCh38
rs770903929 Chr.2: 200818733-200818733 on Build GRCh38
rs763312523 Chr.2: 200818823-200818823 on Build GRCh38
rs774640795 Chr.2: 200818781-200818782 on Build GRCh38
rs754502750 Chr.2: 200818909-200818909 on Build GRCh38
rs753308303 Chr.2: 200818889-200818889 on Build GRCh38
rs927883989 Chr.2: 200818815-200818815 on Build GRCh38
rs1044963508 Chr.2: 200818812-200818812 on Build GRCh38
rs865908843 Chr.2: 200818856-200818856 on Build GRCh38
rs773413596 Chr.2: 200818771-200818771 on Build GRCh38
rs371919876 Chr.2: 200818729-200818729 on Build GRCh38
rs372461709 Chr.2: 200818731-200818731 on Build GRCh38
rs947944517 Chr.2: 200818743-200818743 on Build GRCh38
rs759729287 Chr.2: 200818814-200818814 on Build GRCh38
rs753157996 Chr.2: 200818821-200818821 on Build GRCh38
rs548326048 Chr.2: 200818912-200818912 on Build GRCh38
rs199729181 Chr.2: 200818732-200818732 on Build GRCh38
rs746868014 Chr.2: 200818927-200818927 on Build GRCh38
rs146199590 Chr.2: 200818911-200818911 on Build GRCh38
rs752022036 Chr.2: 200818868-200818868 on Build GRCh38
rs768554108 Chr.2: 200818746-200818746 on Build GRCh38
rs761428764 Chr.2: 200818918-200818918 on Build GRCh38
rs141649308 Chr.2: 200818769-200818769 on Build GRCh38
rs772443127 Chr.2: 200818863-200818864 on Build GRCh38
rs565196210 Chr.2: 200818728-200818728 on Build GRCh38

View Full Product Details