Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127892409-127892670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLRN3
Entrez Gene ID
119467
Gene Name
clarin 3
Gene Aliases
TMEM12, USH3AL1
UniGene
Hs.242014
Gene Chromosome Location
Chr.10: 127877841-127892947 on Build GRCh38
Transcripts
NM_152311.3 XM_011539274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5635025 c.218A>G p.K73R Chr.10: 127892567-127892567 on Build GRCh38
3806727 c.154A>G p.T52A Chr.10: 127892631-127892631 on Build GRCh38
4619942 c.187G>A p.E63K Chr.10: 127892598-127892598 on Build GRCh38
4471093 c.169C>T p.R57C Chr.10: 127892616-127892616 on Build GRCh38
3751607 c.212C>A p.P71Q Chr.10: 127892573-127892573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754104842 Chr.10: 127892616-127892616 on Build GRCh38
rs762524688 Chr.10: 127892525-127892526 on Build GRCh38
rs111836033 Chr.10: 127892633-127892633 on Build GRCh38
rs769642169 Chr.10: 127892605-127892605 on Build GRCh38
rs748695315 Chr.10: 127892583-127892583 on Build GRCh38
rs149266641 Chr.10: 127892615-127892615 on Build GRCh38
rs759755132 Chr.10: 127892618-127892618 on Build GRCh38
rs768895105 Chr.10: 127892509-127892509 on Build GRCh38
rs780924246 Chr.10: 127892600-127892600 on Build GRCh38
rs750637731 Chr.10: 127892559-127892559 on Build GRCh38
rs530269291 Chr.10: 127892453-127892453 on Build GRCh38
rs756384869 Chr.10: 127892560-127892560 on Build GRCh38
rs12267538 Chr.10: 127892464-127892464 on Build GRCh38
rs1004137539 Chr.10: 127892457-127892457 on Build GRCh38
rs775222700 Chr.10: 127892606-127892606 on Build GRCh38
rs370672043 Chr.10: 127892521-127892521 on Build GRCh38
rs765041942 Chr.10: 127892508-127892508 on Build GRCh38
rs754111530 Chr.10: 127892566-127892566 on Build GRCh38
rs908228654 Chr.10: 127892635-127892635 on Build GRCh38
rs777423555 Chr.10: 127892626-127892626 on Build GRCh38
rs150508378 Chr.10: 127892595-127892595 on Build GRCh38
rs1048493536 Chr.10: 127892634-127892634 on Build GRCh38
rs796740073 Chr.10: 127892503-127892503 on Build GRCh38
rs367985220 Chr.10: 127892610-127892610 on Build GRCh38
rs939892579 Chr.10: 127892639-127892639 on Build GRCh38
rs757979351 Chr.10: 127892584-127892584 on Build GRCh38
rs35070529 Chr.10: 127892562-127892562 on Build GRCh38
rs765701577 Chr.10: 127892625-127892625 on Build GRCh38
rs201802645 Chr.10: 127892510-127892510 on Build GRCh38
rs921004163 Chr.10: 127892446-127892446 on Build GRCh38
rs75572194 Chr.10: 127892604-127892604 on Build GRCh38
rs531791079 Chr.10: 127892591-127892591 on Build GRCh38
rs773328252 Chr.10: 127892613-127892613 on Build GRCh38
rs374334834 Chr.10: 127892587-127892587 on Build GRCh38
rs755393004 Chr.10: 127892567-127892567 on Build GRCh38
rs35890579 Chr.10: 127892573-127892573 on Build GRCh38
rs895666053 Chr.10: 127892494-127892494 on Build GRCh38
rs145641755 Chr.10: 127892569-127892569 on Build GRCh38

View Full Product Details