Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45613468-45613715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML2
Entrez Gene ID
24139
Gene Name
echinoderm microtubule associated protein like 2
Gene Aliases
ELP70, EMAP-2, EMAP2
UniGene
Hs.24178
Gene Chromosome Location
Chr.19: 45609396-45645601 on Build GRCh38
Transcripts
NM_001193268.1 NM_001193269.1 NM_012155.2 NR_034098.1 XM_006723125.2 XM_006723126.2 XM_011526694.2 XM_011526695.2 XM_011526698.2 XM_011526700.2 XM_011526701.2 XM_017026550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4946765 c.2135-1G>A p.? Chr.19: 45613672-45613672 on Build GRCh38
4785602 c.2330T>C p.I777T Chr.19: 45613638-45613638 on Build GRCh38
4781763 c.2235C>T p.H745H Chr.19: 45613571-45613571 on Build GRCh38
4851777 c.2218G>T p.D740Y Chr.19: 45613588-45613588 on Build GRCh38
3404390 c.1794C>T p.H598H Chr.19: 45613571-45613571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766544040 Chr.19: 45613604-45613604 on Build GRCh38
rs925845554 Chr.19: 45613607-45613607 on Build GRCh38
rs367980918 Chr.19: 45613543-45613543 on Build GRCh38
rs188496855 Chr.19: 45613555-45613555 on Build GRCh38
rs369947826 Chr.19: 45613606-45613606 on Build GRCh38
rs752653220 Chr.19: 45613684-45613684 on Build GRCh38
rs768074973 Chr.19: 45613560-45613560 on Build GRCh38
rs745601081 Chr.19: 45613624-45613624 on Build GRCh38
rs758689547 Chr.19: 45613510-45613510 on Build GRCh38
rs139893249 Chr.19: 45613649-45613649 on Build GRCh38
rs537364147 Chr.19: 45613558-45613558 on Build GRCh38
rs765633746 Chr.19: 45613587-45613587 on Build GRCh38
rs763326087 Chr.19: 45613593-45613593 on Build GRCh38
rs755057669 Chr.19: 45613611-45613611 on Build GRCh38
rs772014629 Chr.19: 45613626-45613626 on Build GRCh38
rs754334260 Chr.19: 45613491-45613491 on Build GRCh38
rs971185542 Chr.19: 45613679-45613679 on Build GRCh38
rs932421789 Chr.19: 45613496-45613496 on Build GRCh38
rs920908704 Chr.19: 45613683-45613683 on Build GRCh38
rs765572456 Chr.19: 45613501-45613501 on Build GRCh38
rs1025518208 Chr.19: 45613665-45613665 on Build GRCh38
rs772180557 Chr.19: 45613636-45613638 on Build GRCh38
rs372565896 Chr.19: 45613623-45613623 on Build GRCh38
rs376700086 Chr.19: 45613688-45613688 on Build GRCh38
rs766772992 Chr.19: 45613657-45613657 on Build GRCh38
rs747021128 Chr.19: 45613512-45613512 on Build GRCh38
rs556895545 Chr.19: 45613574-45613574 on Build GRCh38
rs763267506 Chr.19: 45613653-45613653 on Build GRCh38
rs144652162 Chr.19: 45613633-45613633 on Build GRCh38
rs778944633 Chr.19: 45613689-45613689 on Build GRCh38
rs755040416 Chr.19: 45613524-45613524 on Build GRCh38
rs762374580 Chr.19: 45613639-45613639 on Build GRCh38
rs759736912 Chr.19: 45613678-45613678 on Build GRCh38
rs745529852 Chr.19: 45613507-45613507 on Build GRCh38
rs1012323345 Chr.19: 45613655-45613655 on Build GRCh38
rs751787095 Chr.19: 45613672-45613672 on Build GRCh38
rs576798996 Chr.19: 45613638-45613638 on Build GRCh38
rs773869032 Chr.19: 45613507-45613507 on Build GRCh38
rs4445998 Chr.19: 45613529-45613529 on Build GRCh38
rs757594055 Chr.19: 45613499-45613499 on Build GRCh38
rs750416002 Chr.19: 45613690-45613690 on Build GRCh38
rs757209617 Chr.19: 45613620-45613620 on Build GRCh38
rs777714421 Chr.19: 45613527-45613527 on Build GRCh38
rs967203926 Chr.19: 45613542-45613542 on Build GRCh38
rs758234710 Chr.19: 45613693-45613693 on Build GRCh38
rs760192491 Chr.19: 45613557-45613557 on Build GRCh38
rs777175792 Chr.19: 45613564-45613564 on Build GRCh38
rs534842289 Chr.19: 45613511-45613511 on Build GRCh38
rs748116326 Chr.19: 45613610-45613610 on Build GRCh38
rs768188753 Chr.19: 45613634-45613634 on Build GRCh38
rs750849410 Chr.19: 45613589-45613589 on Build GRCh38
rs781181360 Chr.19: 45613618-45613618 on Build GRCh38
rs375442814 Chr.19: 45613571-45613571 on Build GRCh38
rs750615251 Chr.19: 45613508-45613508 on Build GRCh38

View Full Product Details