Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 22880018-22879789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP1
Entrez Gene ID
23191
Gene Name
cytoplasmic FMR1 interacting protein 1
Gene Aliases
P140SRA-1, SHYC, SRA-1, SRA1
UniGene
Hs.26704
Gene Chromosome Location
Chr.15: 22869465-22980906 on Build GRCh38
Transcripts
NM_001033028.1 NM_001287810.2 NM_001324119.1 NM_001324120.1 NM_001324122.1 NM_001324123.1 NM_001324124.1 NM_001324125.1 NM_001324126.1 NM_014608.4 XM_011543873.2 XM_011543874.1 XM_011543876.2 XM_017022023.1 XM_017022024.1

View Full Product Details