Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42165208-42165457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKH
Entrez Gene ID
160851
Gene Name
diacylglycerol kinase eta
Gene Aliases
DGKeta
UniGene
Hs.630380
Gene Chromosome Location
Chr.13: 42040036-42256584 on Build GRCh38
Transcripts
NM_001204504.2 NM_001204505.2 NM_001204506.2 NM_001297429.1 NM_152910.5 NM_178009.4 NR_123714.1 NR_123715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4214960 c.536G>A p.S179N Chr.13: 42165419-42165419 on Build GRCh38
7097038 c.958G>T p.G320C Chr.13: 42165433-42165433 on Build GRCh38
1941001 c.944G>A p.S315N Chr.13: 42165419-42165419 on Build GRCh38
7287021 c.495T>C p.G165G Chr.13: 42165378-42165378 on Build GRCh38
7093387 c.955G>A p.D319N Chr.13: 42165430-42165430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779150570 Chr.13: 42165364-42165364 on Build GRCh38
rs781532969 Chr.13: 42165284-42165284 on Build GRCh38
rs976902308 Chr.13: 42165264-42165264 on Build GRCh38
rs562550662 Chr.13: 42165430-42165430 on Build GRCh38
rs141975148 Chr.13: 42165400-42165400 on Build GRCh38
rs1029662362 Chr.13: 42165268-42165268 on Build GRCh38
rs760144949 Chr.13: 42165386-42165386 on Build GRCh38
rs867761822 Chr.13: 42165327-42165327 on Build GRCh38
rs372920372 Chr.13: 42165381-42165381 on Build GRCh38
rs752425859 Chr.13: 42165416-42165416 on Build GRCh38
rs762859675 Chr.13: 42165424-42165424 on Build GRCh38
rs750985985 Chr.13: 42165412-42165412 on Build GRCh38
rs560151754 Chr.13: 42165286-42165286 on Build GRCh38
rs144691483 Chr.13: 42165289-42165289 on Build GRCh38
rs781538723 Chr.13: 42165306-42165306 on Build GRCh38
rs936036829 Chr.13: 42165423-42165423 on Build GRCh38
rs748611706 Chr.13: 42165372-42165372 on Build GRCh38
rs780669381 Chr.13: 42165323-42165323 on Build GRCh38
rs769008336 Chr.13: 42165342-42165342 on Build GRCh38
rs1054653950 Chr.13: 42165277-42165277 on Build GRCh38
rs148496237 Chr.13: 42165394-42165394 on Build GRCh38
rs374539521 Chr.13: 42165429-42165429 on Build GRCh38
rs745924263 Chr.13: 42165313-42165313 on Build GRCh38
rs756305639 Chr.13: 42165320-42165320 on Build GRCh38
rs771610944 Chr.13: 42165376-42165376 on Build GRCh38
rs765012150 Chr.13: 42165408-42165408 on Build GRCh38
rs759478548 Chr.13: 42165406-42165406 on Build GRCh38
rs372307402 Chr.13: 42165293-42165293 on Build GRCh38
rs749837381 Chr.13: 42165338-42165338 on Build GRCh38

View Full Product Details