Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79080998-79081223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENGASE
Entrez Gene ID
64772
Gene Name
endo-beta-N-acetylglucosaminidase
Gene Aliases
-
UniGene
Hs.29288
Gene Chromosome Location
Chr.17: 79074812-79088603 on Build GRCh38
Transcripts
NM_001042573.2 XM_006722017.3 XM_006722019.3 XM_017024961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
707766 c.853G>A p.E285K Chr.17: 79081054-79081054 on Build GRCh38
6317520 c.820G>T p.V274L Chr.17: 79081021-79081021 on Build GRCh38
4794918 c.823G>A p.V275M Chr.17: 79081024-79081024 on Build GRCh38
4794919 c.265G>A p.V89M Chr.17: 79081024-79081024 on Build GRCh38
4070258 c.863A>T p.Q288L Chr.17: 79081064-79081064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996849489 Chr.17: 79081196-79081196 on Build GRCh38
rs990719734 Chr.17: 79081150-79081150 on Build GRCh38
rs772343762 Chr.17: 79081020-79081020 on Build GRCh38
rs1055260098 Chr.17: 79081046-79081046 on Build GRCh38
rs58652925 Chr.17: 79081088-79081088 on Build GRCh38
rs747813884 Chr.17: 79081108-79081108 on Build GRCh38
rs552537922 Chr.17: 79081146-79081146 on Build GRCh38
rs761531574 Chr.17: 79081044-79081044 on Build GRCh38
rs772716577 Chr.17: 79081123-79081123 on Build GRCh38
rs913362837 Chr.17: 79081155-79081155 on Build GRCh38
rs373182387 Chr.17: 79081074-79081074 on Build GRCh38
rs547678422 Chr.17: 79081054-79081054 on Build GRCh38
rs200512199 Chr.17: 79081122-79081122 on Build GRCh38
rs765933875 Chr.17: 79081064-79081064 on Build GRCh38
rs201348547 Chr.17: 79081091-79081091 on Build GRCh38
rs530151741 Chr.17: 79081112-79081112 on Build GRCh38
rs758330250 Chr.17: 79081062-79081062 on Build GRCh38
rs775696117 Chr.17: 79081021-79081021 on Build GRCh38
rs760777164 Chr.17: 79081032-79081032 on Build GRCh38
rs901294001 Chr.17: 79081188-79081188 on Build GRCh38
rs911281755 Chr.17: 79081144-79081144 on Build GRCh38
rs886911205 Chr.17: 79081089-79081089 on Build GRCh38
rs757601617 Chr.17: 79081114-79081114 on Build GRCh38
rs777991338 Chr.17: 79081094-79081095 on Build GRCh38
rs776869235 Chr.17: 79081043-79081043 on Build GRCh38
rs754419091 Chr.17: 79081084-79081084 on Build GRCh38
rs764413417 Chr.17: 79081037-79081037 on Build GRCh38
rs1055696779 Chr.17: 79081087-79081103 on Build GRCh38
rs73398098 Chr.17: 79081140-79081140 on Build GRCh38
rs779331050 Chr.17: 79081119-79081119 on Build GRCh38
rs549837539 Chr.17: 79081116-79081116 on Build GRCh38
rs767265483 Chr.17: 79081086-79081086 on Build GRCh38
rs934246821 Chr.17: 79081138-79081138 on Build GRCh38
rs1016813134 Chr.17: 79081077-79081077 on Build GRCh38
rs765063072 Chr.17: 79081053-79081053 on Build GRCh38
rs565837030 Chr.17: 79081184-79081184 on Build GRCh38
rs1028290217 Chr.17: 79081201-79081201 on Build GRCh38

View Full Product Details