Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49514244-49514487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGRT
Entrez Gene ID
2217
Gene Name
Fc fragment of IgG receptor and transporter
Gene Aliases
FCRN, alpha-chain
UniGene
Hs.111903
Gene Chromosome Location
Chr.19: 49512279-49526428 on Build GRCh38
Transcripts
NM_001136019.2 NM_004107.4 XM_005258657.1 XM_011526627.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5445542 c.435C>G p.L145L Chr.19: 49514320-49514320 on Build GRCh38
713404 c.513C>T p.A171A Chr.19: 49514398-49514398 on Build GRCh38
5435336 c.390C>T p.T130T Chr.19: 49514275-49514275 on Build GRCh38
6406364 c.581G>A p.R194H Chr.19: 49514466-49514466 on Build GRCh38
7238327 c.429C>T p.F143F Chr.19: 49514314-49514314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376030037 Chr.19: 49514437-49514437 on Build GRCh38
rs772294645 Chr.19: 49514425-49514427 on Build GRCh38
rs766823478 Chr.19: 49514462-49514462 on Build GRCh38
rs75776802 Chr.19: 49514339-49514339 on Build GRCh38
rs965231176 Chr.19: 49514327-49514327 on Build GRCh38
rs374093601 Chr.19: 49514357-49514357 on Build GRCh38
rs777861971 Chr.19: 49514282-49514282 on Build GRCh38
rs763553495 Chr.19: 49514461-49514461 on Build GRCh38
rs759988303 Chr.19: 49514465-49514465 on Build GRCh38
rs2878342 Chr.19: 49514467-49514467 on Build GRCh38
rs773834526 Chr.19: 49514342-49514342 on Build GRCh38
rs976629364 Chr.19: 49514422-49514422 on Build GRCh38
rs200637204 Chr.19: 49514425-49514425 on Build GRCh38
rs760999604 Chr.19: 49514326-49514326 on Build GRCh38
rs766812943 Chr.19: 49514363-49514363 on Build GRCh38
rs373482955 Chr.19: 49514432-49514432 on Build GRCh38
rs150420714 Chr.19: 49514281-49514281 on Build GRCh38
rs921214557 Chr.19: 49514387-49514387 on Build GRCh38
rs767296631 Chr.19: 49514394-49514394 on Build GRCh38
rs777412760 Chr.19: 49514401-49514401 on Build GRCh38
rs759996121 Chr.19: 49514452-49514452 on Build GRCh38
rs776785568 Chr.19: 49514328-49514328 on Build GRCh38
rs548660464 Chr.19: 49514331-49514331 on Build GRCh38
rs1036479769 Chr.19: 49514434-49514434 on Build GRCh38
rs749180876 Chr.19: 49514284-49514284 on Build GRCh38
rs751836833 Chr.19: 49514368-49514368 on Build GRCh38
rs145251123 Chr.19: 49514444-49514444 on Build GRCh38
rs200294618 Chr.19: 49514439-49514439 on Build GRCh38
rs753321487 Chr.19: 49514468-49514468 on Build GRCh38
rs774868778 Chr.19: 49514464-49514464 on Build GRCh38
rs554770888 Chr.19: 49514433-49514433 on Build GRCh38
rs140957263 Chr.19: 49514466-49514466 on Build GRCh38
rs985504039 Chr.19: 49514272-49514272 on Build GRCh38
rs754562838 Chr.19: 49514275-49514275 on Build GRCh38
rs932442681 Chr.19: 49514389-49514389 on Build GRCh38
rs369173603 Chr.19: 49514395-49514395 on Build GRCh38
rs751281836 Chr.19: 49514274-49514274 on Build GRCh38
rs530230990 Chr.19: 49514288-49514288 on Build GRCh38
rs773997430 Chr.19: 49514315-49514315 on Build GRCh38
rs755325686 Chr.19: 49514377-49514377 on Build GRCh38
rs749298979 Chr.19: 49514413-49514413 on Build GRCh38
rs780275784 Chr.19: 49514270-49514270 on Build GRCh38
rs140458678 Chr.19: 49514280-49514280 on Build GRCh38
rs1050951160 Chr.19: 49514417-49514417 on Build GRCh38
rs765233786 Chr.19: 49514337-49514337 on Build GRCh38

View Full Product Details