Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71224444-71224669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJB1
Entrez Gene ID
2705
Gene Name
gap junction protein beta 1
Gene Aliases
CMTX, CMTX1, CX32
UniGene
Hs.333303
Gene Chromosome Location
Chr.X: 71215212-71225215 on Build GRCh38
Transcripts
NM_000166.5 NM_001097642.2 XM_011530907.2 XM_017029408.1

SNP Detail

SNP ID Chromosome Location
rs762776007 Chr.X: 71224594-71224594 on Build GRCh38
rs771578720 Chr.X: 71224617-71224617 on Build GRCh38
rs1042771 Chr.X: 71224589-71224589 on Build GRCh38
rs765128969 Chr.X: 71224480-71224480 on Build GRCh38
rs749605877 Chr.X: 71224531-71224531 on Build GRCh38
rs770941057 Chr.X: 71224602-71224602 on Build GRCh38
rs374321066 Chr.X: 71224542-71224542 on Build GRCh38
rs780427727 Chr.X: 71224530-71224530 on Build GRCh38
rs769121326 Chr.X: 71224578-71224578 on Build GRCh38
rs769526542 Chr.X: 71224588-71224588 on Build GRCh38
rs113014279 Chr.X: 71224593-71224593 on Build GRCh38
rs866253945 Chr.X: 71224549-71224549 on Build GRCh38
rs750620543 Chr.X: 71224514-71224514 on Build GRCh38
rs751486192 Chr.X: 71224499-71224499 on Build GRCh38
rs774283188 Chr.X: 71224631-71224631 on Build GRCh38
rs587777879 Chr.X: 71224497-71224497 on Build GRCh38
rs1017639258 Chr.X: 71224543-71224543 on Build GRCh38
rs770362340 Chr.X: 71224558-71224558 on Build GRCh38
rs779327968 Chr.X: 71224550-71224550 on Build GRCh38
rs1055605737 Chr.X: 71224566-71224566 on Build GRCh38
rs1032823915 Chr.X: 71224595-71224595 on Build GRCh38
rs867277984 Chr.X: 71224505-71224505 on Build GRCh38
rs763991026 Chr.X: 71224498-71224498 on Build GRCh38
rs781315897 Chr.X: 71224501-71224501 on Build GRCh38
rs752501254 Chr.X: 71224491-71224491 on Build GRCh38
rs763949039 Chr.X: 71224598-71224598 on Build GRCh38
rs745547989 Chr.X: 71224581-71224581 on Build GRCh38
rs756258267 Chr.X: 71224518-71224518 on Build GRCh38
rs748676943 Chr.X: 71224556-71224556 on Build GRCh38
rs748095080 Chr.X: 71224537-71224537 on Build GRCh38
rs774210122 Chr.X: 71224600-71224600 on Build GRCh38

View Full Product Details