Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74572231-74572502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH3
Entrez Gene ID
127254
Gene Name
glutamate rich 3
Gene Aliases
C1orf173
UniGene
Hs.531182
Gene Chromosome Location
Chr.1: 74568111-74674343 on Build GRCh38
Transcripts
NM_001002912.4 XM_017000274.1 XM_017000275.1 XM_017000276.1 XM_017000277.1 XM_017000278.1 XM_017000279.1 XM_017000280.1 XM_017000281.1 XM_017000282.1 XM_017000283.1 XM_017000284.1 XM_017000285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3772082 c.3380A>G p.E1127G Chr.1: 74572330-74572330 on Build GRCh38
536094 c.3298G>A p.A1100T Chr.1: 74572412-74572412 on Build GRCh38
336645 c.3208C>G p.L1070V Chr.1: 74572502-74572502 on Build GRCh38
7163111 c.3302A>T p.E1101V Chr.1: 74572408-74572408 on Build GRCh38
6311329 c.3256C>A p.L1086I Chr.1: 74572454-74572454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766264276 Chr.1: 74572361-74572361 on Build GRCh38
rs779838426 Chr.1: 74572326-74572346 on Build GRCh38
rs761780276 Chr.1: 74572258-74572258 on Build GRCh38
rs752184955 Chr.1: 74572302-74572302 on Build GRCh38
rs200770042 Chr.1: 74572407-74572407 on Build GRCh38
rs766061112 Chr.1: 74572284-74572284 on Build GRCh38
rs940653335 Chr.1: 74572444-74572444 on Build GRCh38
rs145817694 Chr.1: 74572428-74572428 on Build GRCh38
rs1055608153 Chr.1: 74572418-74572418 on Build GRCh38
rs186027985 Chr.1: 74572385-74572385 on Build GRCh38
rs780172399 Chr.1: 74572410-74572410 on Build GRCh38
rs781504771 Chr.1: 74572394-74572394 on Build GRCh38
rs1029216646 Chr.1: 74572277-74572277 on Build GRCh38
rs140321323 Chr.1: 74572366-74572366 on Build GRCh38
rs754644777 Chr.1: 74572285-74572285 on Build GRCh38
rs754937800 Chr.1: 74572389-74572389 on Build GRCh38
rs758913679 Chr.1: 74572308-74572308 on Build GRCh38
rs757831599 Chr.1: 74572391-74572391 on Build GRCh38
rs931202045 Chr.1: 74572425-74572425 on Build GRCh38
rs769950814 Chr.1: 74572326-74572326 on Build GRCh38
rs981224325 Chr.1: 74572403-74572403 on Build GRCh38
rs775022350 Chr.1: 74572455-74572455 on Build GRCh38
rs781149287 Chr.1: 74572391-74572392 on Build GRCh38
rs373888542 Chr.1: 74572412-74572412 on Build GRCh38
rs778477713 Chr.1: 74572292-74572292 on Build GRCh38
rs760487344 Chr.1: 74572352-74572352 on Build GRCh38
rs139996120 Chr.1: 74572351-74572351 on Build GRCh38
rs764901117 Chr.1: 74572445-74572445 on Build GRCh38
rs144966818 Chr.1: 74572384-74572384 on Build GRCh38
rs144055915 Chr.1: 74572464-74572464 on Build GRCh38
rs370779572 Chr.1: 74572343-74572343 on Build GRCh38
rs200242395 Chr.1: 74572379-74572379 on Build GRCh38
rs190095556 Chr.1: 74572401-74572401 on Build GRCh38
rs564254097 Chr.1: 74572411-74572411 on Build GRCh38
rs767405824 Chr.1: 74572259-74572259 on Build GRCh38
rs757852992 Chr.1: 74572376-74572376 on Build GRCh38
rs775619326 Chr.1: 74572337-74572337 on Build GRCh38
rs781598691 Chr.1: 74572316-74572316 on Build GRCh38
rs376202548 Chr.1: 74572320-74572320 on Build GRCh38
rs182380383 Chr.1: 74572414-74572414 on Build GRCh38
rs757739631 Chr.1: 74572465-74572465 on Build GRCh38
rs533198887 Chr.1: 74572310-74572310 on Build GRCh38
rs752199834 Chr.1: 74572368-74572368 on Build GRCh38
rs574677549 Chr.1: 74572276-74572276 on Build GRCh38
rs759269285 Chr.1: 74572432-74572432 on Build GRCh38
rs763594793 Chr.1: 74572462-74572462 on Build GRCh38
rs201136666 Chr.1: 74572442-74572442 on Build GRCh38
rs989801836 Chr.1: 74572356-74572356 on Build GRCh38
rs139688086 Chr.1: 74572271-74572271 on Build GRCh38
rs762966601 Chr.1: 74572345-74572345 on Build GRCh38
rs866558257 Chr.1: 74572325-74572325 on Build GRCh38
rs776460660 Chr.1: 74572430-74572430 on Build GRCh38
rs746938882 Chr.1: 74572419-74572419 on Build GRCh38
rs746755928 Chr.1: 74572387-74572388 on Build GRCh38

View Full Product Details