Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45120407-45120680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOXA1
Entrez Gene ID
90527
Gene Name
dual oxidase maturation factor 1
Gene Aliases
NIP, NUMBIP, mol
UniGene
Hs.356664
Gene Chromosome Location
Chr.15: 45117366-45129938 on Build GRCh38
Transcripts
NM_001276264.1 NM_001276265.1 NM_001276266.1 NM_001276267.1 NM_001276268.1 NM_144565.3 XM_006720743.3 XM_006720745.1 XM_006720746.1 XM_006720747.3 XM_006720749.1 XM_006720751.3 XM_006720752.3 XM_006720754.1 XM_006720755.1 XM_011522181.2 XM_011522182.1 XM_011522183.2 XM_011522184.1 XM_011522185.1 XM_011522186.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
470712 c.520C>T p.R174C Chr.15: 45120626-45120626 on Build GRCh38
4779713 c.466T>C p.Y156H Chr.15: 45120680-45120680 on Build GRCh38
470711 c.520_521CG>TT p.R174>? Chr.15: 45120625-45120626 on Build GRCh38
4251535 c.528G>A p.A176A Chr.15: 45120618-45120618 on Build GRCh38
1708104 c.497C>T p.P166L Chr.15: 45120649-45120649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147797857 Chr.15: 45120634-45120635 on Build GRCh38
rs976700740 Chr.15: 45120609-45120609 on Build GRCh38
rs374208931 Chr.15: 45120581-45120581 on Build GRCh38
rs746258066 Chr.15: 45120619-45120619 on Build GRCh38
rs376672007 Chr.15: 45120601-45120601 on Build GRCh38
rs780875523 Chr.15: 45120591-45120591 on Build GRCh38
rs1000747176 Chr.15: 45120505-45120505 on Build GRCh38
rs957831683 Chr.15: 45120537-45120537 on Build GRCh38
rs760939470 Chr.15: 45120635-45120635 on Build GRCh38
rs752365433 Chr.15: 45120594-45120594 on Build GRCh38
rs76617949 Chr.15: 45120637-45120637 on Build GRCh38
rs187988297 Chr.15: 45120481-45120481 on Build GRCh38
rs766330670 Chr.15: 45120642-45120642 on Build GRCh38
rs763694078 Chr.15: 45120636-45120636 on Build GRCh38
rs528322709 Chr.15: 45120519-45120519 on Build GRCh38
rs559554507 Chr.15: 45120504-45120504 on Build GRCh38
rs574036594 Chr.15: 45120462-45120462 on Build GRCh38
rs368996883 Chr.15: 45120572-45120572 on Build GRCh38
rs766238678 Chr.15: 45120576-45120576 on Build GRCh38
rs1016059230 Chr.15: 45120598-45120598 on Build GRCh38
rs987981633 Chr.15: 45120562-45120562 on Build GRCh38
rs775613764 Chr.15: 45120634-45120634 on Build GRCh38
rs777321207 Chr.15: 45120606-45120606 on Build GRCh38
rs779521829 Chr.15: 45120618-45120618 on Build GRCh38
rs774253631 Chr.15: 45120643-45120643 on Build GRCh38
rs182466172 Chr.15: 45120440-45120440 on Build GRCh38
rs569682046 Chr.15: 45120436-45120436 on Build GRCh38
rs772455041 Chr.15: 45120545-45120545 on Build GRCh38
rs946139552 Chr.15: 45120454-45120454 on Build GRCh38
rs771565274 Chr.15: 45120617-45120617 on Build GRCh38
rs535394171 Chr.15: 45120477-45120477 on Build GRCh38
rs775169480 Chr.15: 45120498-45120498 on Build GRCh38
rs147223934 Chr.15: 45120625-45120625 on Build GRCh38
rs764201573 Chr.15: 45120568-45120568 on Build GRCh38
rs748684177 Chr.15: 45120613-45120613 on Build GRCh38
rs1001944739 Chr.15: 45120515-45120515 on Build GRCh38
rs1024633431 Chr.15: 45120444-45120444 on Build GRCh38
rs12916804 Chr.15: 45120602-45120602 on Build GRCh38
rs761760347 Chr.15: 45120641-45120641 on Build GRCh38
rs374636586 Chr.15: 45120573-45120573 on Build GRCh38
rs969490464 Chr.15: 45120452-45120452 on Build GRCh38
rs760715555 Chr.15: 45120552-45120552 on Build GRCh38
rs547614157 Chr.15: 45120533-45120533 on Build GRCh38
rs1050504979 Chr.15: 45120630-45120630 on Build GRCh38
rs148263159 Chr.15: 45120649-45120649 on Build GRCh38

View Full Product Details