Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1215794-1216031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBP1
Entrez Gene ID
1487
Gene Name
C-terminal binding protein 1
Gene Aliases
BARS
UniGene
Hs.208597
Gene Chromosome Location
Chr.4: 1211440-1250329 on Build GRCh38
Transcripts
NM_001012614.1 NM_001328.2 XM_005272261.3 XM_005272263.4 XM_011513395.2 XM_011513396.2 XM_011513397.2 XM_017007762.1 XM_017007763.1 XM_017007764.1 XM_017007765.1 XM_017007766.1 XM_017007767.1 NR_104331.1

Gene Symbol
CTBP1-AS
Entrez Gene ID
285463
Gene Name
CTBP1 antisense RNA
Gene Aliases
PCAT10
UniGene
Hs.707614
Gene Chromosome Location
Chr.4: 1210120-1218591 on Build GRCh38
Transcripts
NM_001012614.1 NM_001328.2 XM_005272261.3 XM_005272263.4 XM_011513395.2 XM_011513396.2 XM_011513397.2 XM_017007762.1 XM_017007763.1 XM_017007764.1 XM_017007765.1 XM_017007766.1 XM_017007767.1 NR_104331.1

SNP Detail

SNP ID Chromosome Location
rs908959156 Chr.4: 1215879-1215879 on Build GRCh38
rs923204867 Chr.4: 1215922-1215922 on Build GRCh38
rs949804140 Chr.4: 1215889-1215889 on Build GRCh38
rs73793118 Chr.4: 1215937-1215937 on Build GRCh38
rs991247718 Chr.4: 1215877-1215877 on Build GRCh38
rs763104103 Chr.4: 1215961-1215961 on Build GRCh38
rs73793116 Chr.4: 1215900-1215900 on Build GRCh38
rs774579302 Chr.4: 1215978-1215978 on Build GRCh38
rs147464431 Chr.4: 1216009-1216009 on Build GRCh38
rs964348198 Chr.4: 1215885-1215885 on Build GRCh38
rs770094644 Chr.4: 1215955-1215955 on Build GRCh38
rs143129857 Chr.4: 1216000-1216000 on Build GRCh38
rs763847829 Chr.4: 1216011-1216011 on Build GRCh38
rs771096968 Chr.4: 1215963-1215963 on Build GRCh38
rs542556923 Chr.4: 1215851-1215851 on Build GRCh38
rs769161413 Chr.4: 1215940-1215940 on Build GRCh38
rs375999937 Chr.4: 1215954-1215954 on Build GRCh38
rs952974092 Chr.4: 1215869-1215869 on Build GRCh38
rs759489298 Chr.4: 1215982-1215982 on Build GRCh38
rs934564733 Chr.4: 1215912-1215912 on Build GRCh38
rs781704682 Chr.4: 1215951-1215951 on Build GRCh38
rs773460347 Chr.4: 1215958-1215958 on Build GRCh38
rs767142473 Chr.4: 1215987-1215987 on Build GRCh38
rs778946658 Chr.4: 1215844-1215844 on Build GRCh38
rs972337626 Chr.4: 1215856-1215856 on Build GRCh38
rs767549924 Chr.4: 1215999-1215999 on Build GRCh38
rs753431024 Chr.4: 1215820-1215820 on Build GRCh38
rs141426450 Chr.4: 1215875-1215875 on Build GRCh38
rs561278483 Chr.4: 1215862-1215862 on Build GRCh38
rs1053424484 Chr.4: 1215939-1215939 on Build GRCh38

View Full Product Details