Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95552759-95553018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN10
Entrez Gene ID
9071
Gene Name
claudin 10
Gene Aliases
CPETRL3, OSP-L
UniGene
Hs.534377
Gene Chromosome Location
Chr.13: 95433588-95579756 on Build GRCh38
Transcripts
NM_001160100.1 NM_006984.4 NM_182848.3 XM_011521134.2 XM_017020843.1 XM_017020844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1368064 c.28G>A p.A10T Chr.13: 95552781-95552781 on Build GRCh38
1368065 c.94G>T p.V32L Chr.13: 95552847-95552847 on Build GRCh38
4439119 c.127G>A p.A43T Chr.13: 95552880-95552880 on Build GRCh38
1152673 c.219C>G p.D73E Chr.13: 95552972-95552972 on Build GRCh38
6561744 c.178G>A p.V60I Chr.13: 95552931-95552931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906216130 Chr.13: 95552922-95552922 on Build GRCh38
rs1014516986 Chr.13: 95552809-95552809 on Build GRCh38
rs762701437 Chr.13: 95552912-95552912 on Build GRCh38
rs932217780 Chr.13: 95552853-95552853 on Build GRCh38
rs546191968 Chr.13: 95552954-95552954 on Build GRCh38
rs779532016 Chr.13: 95552982-95552982 on Build GRCh38
rs757681796 Chr.13: 95552921-95552921 on Build GRCh38
rs200066195 Chr.13: 95552828-95552828 on Build GRCh38
rs368313176 Chr.13: 95552940-95552940 on Build GRCh38
rs576857690 Chr.13: 95552953-95552953 on Build GRCh38
rs757992913 Chr.13: 95552801-95552801 on Build GRCh38
rs773460504 Chr.13: 95552861-95552861 on Build GRCh38
rs868569022 Chr.13: 95552903-95552903 on Build GRCh38
rs115599668 Chr.13: 95552792-95552792 on Build GRCh38
rs534072709 Chr.13: 95552811-95552811 on Build GRCh38
rs764942392 Chr.13: 95552790-95552790 on Build GRCh38
rs756783148 Chr.13: 95552812-95552812 on Build GRCh38
rs753799088 Chr.13: 95552781-95552781 on Build GRCh38
rs748713153 Chr.13: 95552984-95552984 on Build GRCh38
rs907066795 Chr.13: 95552993-95552993 on Build GRCh38
rs766998645 Chr.13: 95552946-95552946 on Build GRCh38
rs747186983 Chr.13: 95552996-95552996 on Build GRCh38
rs772363407 Chr.13: 95552860-95552860 on Build GRCh38
rs370647506 Chr.13: 95552829-95552829 on Build GRCh38
rs779172671 Chr.13: 95552998-95552998 on Build GRCh38
rs763834792 Chr.13: 95552808-95552808 on Build GRCh38
rs563014813 Chr.13: 95552979-95552979 on Build GRCh38
rs771037194 Chr.13: 95552881-95552881 on Build GRCh38
rs201714839 Chr.13: 95552900-95552900 on Build GRCh38
rs374282781 Chr.13: 95552938-95552938 on Build GRCh38
rs747253181 Chr.13: 95552880-95552880 on Build GRCh38
rs759535516 Chr.13: 95552788-95552788 on Build GRCh38
rs763856292 Chr.13: 95552916-95552916 on Build GRCh38
rs758776274 Chr.13: 95552990-95552990 on Build GRCh38
rs1010607066 Chr.13: 95552972-95552972 on Build GRCh38
rs776695497 Chr.13: 95552884-95552884 on Build GRCh38
rs778064352 Chr.13: 95552995-95552995 on Build GRCh38
rs759408749 Chr.13: 95552897-95552897 on Build GRCh38
rs757685709 Chr.13: 95552952-95552952 on Build GRCh38
rs748553948 Chr.13: 95552858-95552858 on Build GRCh38
rs142907955 Chr.13: 95552951-95552951 on Build GRCh38
rs755503837 Chr.13: 95552959-95552959 on Build GRCh38
rs779373596 Chr.13: 95552844-95552844 on Build GRCh38

View Full Product Details