Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77442473-77442746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIPAS39
Entrez Gene ID
63894
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
Gene Aliases
C14orf133, SPE-39, SPE39, VIPAR, VPS16B, hSPE-39
UniGene
Hs.16157
Gene Chromosome Location
Chr.14: 77426675-77457640 on Build GRCh38
Transcripts
NM_001193314.1 NM_001193315.1 NM_001193316.1 NM_001193317.1 NM_022067.3 XM_011537066.1 XM_017021579.1 XM_017021580.1 XM_017021581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4803223 c.659G>A p.R220Q Chr.14: 77442635-77442635 on Build GRCh38
3782850 c.658C>T p.R220* Chr.14: 77442636-77442636 on Build GRCh38
3769516 c.644G>A p.R215Q Chr.14: 77442650-77442650 on Build GRCh38
699292 c.632-1G>T p.? Chr.14: 77442663-77442663 on Build GRCh38
5559056 c.728T>C p.L243P Chr.14: 77442566-77442566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770441661 Chr.14: 77442511-77442511 on Build GRCh38
rs377615868 Chr.14: 77442651-77442651 on Build GRCh38
rs577625484 Chr.14: 77442520-77442520 on Build GRCh38
rs762911709 Chr.14: 77442546-77442546 on Build GRCh38
rs765276366 Chr.14: 77442527-77442527 on Build GRCh38
rs751402602 Chr.14: 77442689-77442689 on Build GRCh38
rs757522417 Chr.14: 77442586-77442586 on Build GRCh38
rs201486271 Chr.14: 77442675-77442675 on Build GRCh38
rs200370925 Chr.14: 77442636-77442636 on Build GRCh38
rs751882120 Chr.14: 77442567-77442567 on Build GRCh38
rs764078341 Chr.14: 77442686-77442686 on Build GRCh38
rs370850533 Chr.14: 77442671-77442671 on Build GRCh38
rs761448041 Chr.14: 77442694-77442694 on Build GRCh38
rs946937885 Chr.14: 77442627-77442627 on Build GRCh38
rs780603626 Chr.14: 77442597-77442597 on Build GRCh38
rs373783479 Chr.14: 77442601-77442601 on Build GRCh38
rs775381413 Chr.14: 77442716-77442716 on Build GRCh38
rs1034609076 Chr.14: 77442598-77442598 on Build GRCh38
rs115694495 Chr.14: 77442696-77442696 on Build GRCh38
rs1046696029 Chr.14: 77442679-77442679 on Build GRCh38
rs370604803 Chr.14: 77442599-77442599 on Build GRCh38
rs748418108 Chr.14: 77442650-77442650 on Build GRCh38
rs758900238 Chr.14: 77442526-77442526 on Build GRCh38
rs939511004 Chr.14: 77442658-77442658 on Build GRCh38
rs560655868 Chr.14: 77442519-77442519 on Build GRCh38
rs750523421 Chr.14: 77442589-77442589 on Build GRCh38
rs769057172 Chr.14: 77442635-77442635 on Build GRCh38
rs756090677 Chr.14: 77442593-77442593 on Build GRCh38
rs546494893 Chr.14: 77442552-77442552 on Build GRCh38
rs752664898 Chr.14: 77442543-77442543 on Build GRCh38
rs142501624 Chr.14: 77442676-77442676 on Build GRCh38
rs781232441 Chr.14: 77442588-77442588 on Build GRCh38
rs749761450 Chr.14: 77442603-77442603 on Build GRCh38

View Full Product Details