Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162187968-162188235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAP
Entrez Gene ID
2191
Gene Name
fibroblast activation protein alpha
Gene Aliases
DPPIV, FAPA, SIMP
UniGene
Hs.654370
Gene Chromosome Location
Chr.2: 162170684-162243535 on Build GRCh38
Transcripts
NM_001291807.1 NM_004460.3 XM_011510796.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1691220 c.1769G>A p.R590Q Chr.2: 162188214-162188214 on Build GRCh38
1400460 c.1813A>G p.R605G Chr.2: 162188170-162188170 on Build GRCh38
3568509 c.1752C>T p.L584L Chr.2: 162188231-162188231 on Build GRCh38
1400461 c.1804A>G p.T602A Chr.2: 162188179-162188179 on Build GRCh38
1163224 c.1749A>G p.K583K Chr.2: 162188234-162188234 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754414881 Chr.2: 162188207-162188207 on Build GRCh38
rs965363712 Chr.2: 162188094-162188094 on Build GRCh38
rs995805531 Chr.2: 162187999-162187999 on Build GRCh38
rs952206652 Chr.2: 162188035-162188035 on Build GRCh38
rs527951972 Chr.2: 162188178-162188178 on Build GRCh38
rs564688592 Chr.2: 162188045-162188045 on Build GRCh38
rs1051867503 Chr.2: 162188001-162188002 on Build GRCh38
rs750725924 Chr.2: 162188161-162188161 on Build GRCh38
rs980166067 Chr.2: 162188050-162188050 on Build GRCh38
rs1030858847 Chr.2: 162188014-162188014 on Build GRCh38
rs1011630829 Chr.2: 162188029-162188029 on Build GRCh38
rs776936124 Chr.2: 162188125-162188125 on Build GRCh38
rs1018602223 Chr.2: 162188075-162188075 on Build GRCh38
rs999838155 Chr.2: 162188004-162188004 on Build GRCh38
rs957844772 Chr.2: 162188022-162188022 on Build GRCh38
rs980159602 Chr.2: 162188194-162188194 on Build GRCh38
rs532083451 Chr.2: 162188111-162188111 on Build GRCh38
rs976973888 Chr.2: 162188110-162188110 on Build GRCh38
rs139343717 Chr.2: 162188154-162188154 on Build GRCh38
rs956094943 Chr.2: 162188122-162188122 on Build GRCh38
rs150039321 Chr.2: 162188163-162188163 on Build GRCh38
rs752179394 Chr.2: 162188185-162188185 on Build GRCh38
rs868835406 Chr.2: 162188077-162188077 on Build GRCh38
rs534122389 Chr.2: 162188056-162188056 on Build GRCh38
rs764274693 Chr.2: 162188119-162188119 on Build GRCh38
rs928617890 Chr.2: 162188057-162188057 on Build GRCh38
rs765658150 Chr.2: 162188160-162188160 on Build GRCh38
rs571563411 Chr.2: 162188055-162188055 on Build GRCh38
rs960347409 Chr.2: 162188076-162188076 on Build GRCh38

View Full Product Details