Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134661678-134661951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPGM
Entrez Gene ID
669
Gene Name
bisphosphoglycerate mutase
Gene Aliases
DPGM
UniGene
Hs.198365
Gene Chromosome Location
Chr.7: 134646779-134679816 on Build GRCh38
Transcripts
NM_001293085.1 NM_001724.4 NM_199186.2 XM_011516527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3730351 c.184C>T p.R62W Chr.7: 134661691-134661691 on Build GRCh38
2858753 c.219G>T p.E73D Chr.7: 134661726-134661726 on Build GRCh38
6612283 c.269G>A p.R90H Chr.7: 134661776-134661776 on Build GRCh38
4911588 c.321T>C p.H107H Chr.7: 134661828-134661828 on Build GRCh38
6530269 c.315G>C p.L105F Chr.7: 134661822-134661822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026042730 Chr.7: 134661886-134661886 on Build GRCh38
rs756128410 Chr.7: 134661926-134661926 on Build GRCh38
rs372950660 Chr.7: 134661763-134661763 on Build GRCh38
rs755284418 Chr.7: 134661748-134661748 on Build GRCh38
rs144528605 Chr.7: 134661876-134661876 on Build GRCh38
rs775128216 Chr.7: 134661795-134661795 on Build GRCh38
rs750951686 Chr.7: 134661707-134661707 on Build GRCh38
rs901476075 Chr.7: 134661857-134661857 on Build GRCh38
rs190681045 Chr.7: 134661894-134661894 on Build GRCh38
rs768397590 Chr.7: 134661797-134661797 on Build GRCh38
rs770878186 Chr.7: 134661765-134661765 on Build GRCh38
rs762136951 Chr.7: 134661764-134661765 on Build GRCh38
rs1038973044 Chr.7: 134661835-134661835 on Build GRCh38
rs141221644 Chr.7: 134661925-134661925 on Build GRCh38
rs746836937 Chr.7: 134661764-134661764 on Build GRCh38
rs1007481067 Chr.7: 134661828-134661828 on Build GRCh38
rs745809683 Chr.7: 134661788-134661788 on Build GRCh38
rs767426687 Chr.7: 134661886-134661888 on Build GRCh38
rs903716285 Chr.7: 134661822-134661822 on Build GRCh38
rs752985789 Chr.7: 134661908-134661908 on Build GRCh38
rs149329328 Chr.7: 134661851-134661851 on Build GRCh38
rs754001223 Chr.7: 134661740-134661740 on Build GRCh38
rs1043559831 Chr.7: 134661766-134661766 on Build GRCh38
rs553377218 Chr.7: 134661791-134661791 on Build GRCh38
rs374008784 Chr.7: 134661874-134661874 on Build GRCh38
rs781203617 Chr.7: 134661922-134661922 on Build GRCh38
rs573391134 Chr.7: 134661830-134661830 on Build GRCh38
rs781222092 Chr.7: 134661776-134661776 on Build GRCh38
rs762117892 Chr.7: 134661837-134661837 on Build GRCh38
rs774024611 Chr.7: 134661804-134661804 on Build GRCh38
rs756090558 Chr.7: 134661733-134661733 on Build GRCh38
rs780186103 Chr.7: 134661738-134661738 on Build GRCh38
rs546208510 Chr.7: 134661750-134661750 on Build GRCh38
rs201783021 Chr.7: 134661875-134661875 on Build GRCh38
rs773642912 Chr.7: 134661845-134661845 on Build GRCh38
rs543243292 Chr.7: 134661921-134661921 on Build GRCh38
rs765513810 Chr.7: 134661895-134661895 on Build GRCh38
rs376210775 Chr.7: 134661843-134661843 on Build GRCh38
rs548312727 Chr.7: 134661801-134661801 on Build GRCh38
rs370270863 Chr.7: 134661860-134661860 on Build GRCh38
rs121964925 Chr.7: 134661775-134661775 on Build GRCh38

View Full Product Details