Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22070073-22070346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMTN
Entrez Gene ID
2039
Gene Name
dematin actin binding protein
Gene Aliases
DMT, EPB49
UniGene
Hs.106124
Gene Chromosome Location
Chr.8: 22048955-22082527 on Build GRCh38
Transcripts
NM_001114135.4 NM_001114136.2 NM_001114137.3 NM_001114138.2 NM_001114139.3 NM_001302816.2 NM_001302817.2 NM_001323378.1 NM_001323380.1 NM_001323381.1 NM_001323382.1 NM_001323383.1 NM_001323384.1 NM_001323385.1 NM_001323387.1 NM_001323388.1 NM_001323389.1 NM_001323390.1 NM_001323391.1 NM_001323392.1 NM_001323393.1 NM_001323394.1 NM_001323395.1 NM_001323396.1 NM_001323397.1 NM_001323398.1 NM_001323399.1 NM_001323400.1 NM_001323401.1 NM_001978.4 XM_005273432.1 XM_005273433.1 XM_005273442.1 XM_011544435.1 XM_017013189.1 XM_017013190.1 XM_017013191.1 XM_017013192.1 XM_017013193.1 XM_017013194.1 XM_017013195.1 XM_017013196.1 XM_017013197.1 XM_017013198.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4600824 c.505G>A p.E169K Chr.8: 22070235-22070235 on Build GRCh38
3779134 c.490G>A p.E164K Chr.8: 22070220-22070220 on Build GRCh38
7204074 c.532C>A p.P178T Chr.8: 22070262-22070262 on Build GRCh38
4614062 c.582delC p.P196fs*35 Chr.8: 22070312-22070312 on Build GRCh38
3779135 c.490G>A p.E164K Chr.8: 22070220-22070220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749603253 Chr.8: 22070273-22070273 on Build GRCh38
rs74380460 Chr.8: 22070241-22070241 on Build GRCh38
rs373778843 Chr.8: 22070152-22070152 on Build GRCh38
rs776215080 Chr.8: 22070282-22070282 on Build GRCh38
rs1053331534 Chr.8: 22070255-22070255 on Build GRCh38
rs750151796 Chr.8: 22070168-22070168 on Build GRCh38
rs576386712 Chr.8: 22070219-22070219 on Build GRCh38
rs763989834 Chr.8: 22070311-22070312 on Build GRCh38
rs754002326 Chr.8: 22070114-22070114 on Build GRCh38
rs779434697 Chr.8: 22070277-22070277 on Build GRCh38
rs766272009 Chr.8: 22070297-22070297 on Build GRCh38
rs974480257 Chr.8: 22070109-22070109 on Build GRCh38
rs936230491 Chr.8: 22070225-22070225 on Build GRCh38
rs777737281 Chr.8: 22070306-22070306 on Build GRCh38
rs922683892 Chr.8: 22070126-22070126 on Build GRCh38
rs375745249 Chr.8: 22070232-22070232 on Build GRCh38
rs768222260 Chr.8: 22070143-22070143 on Build GRCh38
rs762891978 Chr.8: 22070174-22070174 on Build GRCh38
rs761460751 Chr.8: 22070289-22070289 on Build GRCh38
rs867678554 Chr.8: 22070220-22070220 on Build GRCh38
rs746539661 Chr.8: 22070279-22070279 on Build GRCh38
rs767628320 Chr.8: 22070318-22070318 on Build GRCh38
rs756610356 Chr.8: 22070245-22070245 on Build GRCh38
rs964495465 Chr.8: 22070103-22070103 on Build GRCh38
rs957060881 Chr.8: 22070111-22070111 on Build GRCh38
rs141465910 Chr.8: 22070312-22070312 on Build GRCh38
rs147516712 Chr.8: 22070110-22070110 on Build GRCh38
rs926233015 Chr.8: 22070216-22070216 on Build GRCh38
rs765014831 Chr.8: 22070157-22070157 on Build GRCh38
rs1049935 Chr.8: 22070261-22070261 on Build GRCh38
rs550940280 Chr.8: 22070098-22070099 on Build GRCh38
rs762699437 Chr.8: 22070292-22070292 on Build GRCh38
rs771363592 Chr.8: 22070276-22070276 on Build GRCh38
rs766295458 Chr.8: 22070186-22070186 on Build GRCh38
rs865932605 Chr.8: 22070307-22070307 on Build GRCh38
rs139289658 Chr.8: 22070145-22070145 on Build GRCh38
rs558157418 Chr.8: 22070187-22070187 on Build GRCh38
rs148314902 Chr.8: 22070291-22070291 on Build GRCh38
rs145064007 Chr.8: 22070264-22070264 on Build GRCh38
rs1049934 Chr.8: 22070260-22070260 on Build GRCh38
rs1038462523 Chr.8: 22070249-22070249 on Build GRCh38
rs778282339 Chr.8: 22070246-22070246 on Build GRCh38
rs554264613 Chr.8: 22070106-22070106 on Build GRCh38
rs752754463 Chr.8: 22070321-22070321 on Build GRCh38
rs370045371 Chr.8: 22070234-22070234 on Build GRCh38
rs755057611 Chr.8: 22070205-22070205 on Build GRCh38
rs751494624 Chr.8: 22070309-22070309 on Build GRCh38
rs1016016548 Chr.8: 22070108-22070109 on Build GRCh38
rs376842494 Chr.8: 22070141-22070141 on Build GRCh38
rs761655391 Chr.8: 22070146-22070146 on Build GRCh38
rs760183886 Chr.8: 22070142-22070142 on Build GRCh38

View Full Product Details