Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68422736-68423001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNA2
Entrez Gene ID
1763
Gene Name
DNA replication helicase/nuclease 2
Gene Aliases
DNA2L, hDNA2
UniGene
Hs.532446
Gene Chromosome Location
Chr.10: 68414064-68472521 on Build GRCh38
Transcripts
NM_001080449.2 NR_102264.1 XM_006717680.3 XM_011539417.1 XM_017015799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807526 c.2361C>T p.D787D Chr.10: 68422738-68422738 on Build GRCh38
4015270 c.2479A>C p.T827P Chr.10: 68422878-68422878 on Build GRCh38
84015 c.2312T>C p.I771T Chr.10: 68422787-68422787 on Build GRCh38
3439866 c.2514C>T p.S838S Chr.10: 68422843-68422843 on Build GRCh38
3935141 c.2269G>C p.D757H Chr.10: 68422830-68422830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745984809 Chr.10: 68422885-68422885 on Build GRCh38
rs994781726 Chr.10: 68422813-68422813 on Build GRCh38
rs984360279 Chr.10: 68422820-68422820 on Build GRCh38
rs761641035 Chr.10: 68422778-68422778 on Build GRCh38
rs749588123 Chr.10: 68422903-68422904 on Build GRCh38
rs765854826 Chr.10: 68422808-68422808 on Build GRCh38
rs781772256 Chr.10: 68422763-68422764 on Build GRCh38
rs1027703682 Chr.10: 68422827-68422827 on Build GRCh38
rs149606167 Chr.10: 68422887-68422887 on Build GRCh38
rs868666065 Chr.10: 68422771-68422772 on Build GRCh38
rs764046898 Chr.10: 68422769-68422769 on Build GRCh38
rs866149439 Chr.10: 68422772-68422772 on Build GRCh38
rs774303899 Chr.10: 68422760-68422760 on Build GRCh38
rs776603599 Chr.10: 68422890-68422890 on Build GRCh38
rs761223121 Chr.10: 68422842-68422842 on Build GRCh38
rs920295453 Chr.10: 68422767-68422767 on Build GRCh38
rs538970967 Chr.10: 68422901-68422901 on Build GRCh38
rs764780149 Chr.10: 68422784-68422784 on Build GRCh38
rs756561353 Chr.10: 68422856-68422856 on Build GRCh38
rs755503672 Chr.10: 68422831-68422831 on Build GRCh38
rs751135781 Chr.10: 68422830-68422830 on Build GRCh38
rs774443976 Chr.10: 68422871-68422871 on Build GRCh38
rs761587906 Chr.10: 68422898-68422898 on Build GRCh38
rs10998153 Chr.10: 68422923-68422923 on Build GRCh38
rs753597233 Chr.10: 68422773-68422773 on Build GRCh38
rs374238244 Chr.10: 68422864-68422864 on Build GRCh38
rs141172562 Chr.10: 68422941-68422941 on Build GRCh38
rs780354765 Chr.10: 68422892-68422893 on Build GRCh38
rs1003323248 Chr.10: 68422761-68422761 on Build GRCh38
rs745893364 Chr.10: 68422764-68422764 on Build GRCh38
rs760551270 Chr.10: 68422768-68422768 on Build GRCh38
rs757991769 Chr.10: 68422792-68422792 on Build GRCh38
rs915507777 Chr.10: 68422946-68422946 on Build GRCh38
rs370273531 Chr.10: 68422796-68422796 on Build GRCh38
rs771075304 Chr.10: 68422869-68422869 on Build GRCh38
rs750156670 Chr.10: 68422785-68422785 on Build GRCh38
rs1041709880 Chr.10: 68422966-68422966 on Build GRCh38
rs777857084 Chr.10: 68422857-68422857 on Build GRCh38
rs904555709 Chr.10: 68422976-68422976 on Build GRCh38
rs954417485 Chr.10: 68422910-68422910 on Build GRCh38
rs756274812 Chr.10: 68422772-68422773 on Build GRCh38
rs751092529 Chr.10: 68422841-68422841 on Build GRCh38
rs965820996 Chr.10: 68422872-68422872 on Build GRCh38

View Full Product Details