Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76954140-76954379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHBP
Entrez Gene ID
1393
Gene Name
corticotropin releasing hormone binding protein
Gene Aliases
CRF-BP, CRFBP
UniGene
Hs.115617
Gene Chromosome Location
Chr.5: 76952713-76981143 on Build GRCh38
Transcripts
NM_001882.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3856363 c.333+2T>C p.? Chr.5: 76954188-76954188 on Build GRCh38
6635440 c.306C>T p.I102I Chr.5: 76954159-76954159 on Build GRCh38
1438662 c.322G>A p.D108N Chr.5: 76954175-76954175 on Build GRCh38
1070264 c.321C>T p.G107G Chr.5: 76954174-76954174 on Build GRCh38
6104282 c.295C>A p.Q99K Chr.5: 76954148-76954148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034233872 Chr.5: 76954345-76954346 on Build GRCh38
rs767209029 Chr.5: 76954193-76954193 on Build GRCh38
rs369510749 Chr.5: 76954235-76954235 on Build GRCh38
rs780793331 Chr.5: 76954260-76954260 on Build GRCh38
rs377294117 Chr.5: 76954226-76954226 on Build GRCh38
rs779457908 Chr.5: 76954217-76954217 on Build GRCh38
rs972937457 Chr.5: 76954265-76954265 on Build GRCh38
rs372720988 Chr.5: 76954206-76954206 on Build GRCh38
rs533984056 Chr.5: 76954308-76954308 on Build GRCh38
rs890793369 Chr.5: 76954253-76954253 on Build GRCh38
rs980358034 Chr.5: 76954228-76954228 on Build GRCh38
rs755422412 Chr.5: 76954211-76954211 on Build GRCh38
rs931410179 Chr.5: 76954314-76954314 on Build GRCh38
rs750053690 Chr.5: 76954199-76954199 on Build GRCh38
rs550757067 Chr.5: 76954305-76954305 on Build GRCh38
rs765834486 Chr.5: 76954202-76954202 on Build GRCh38
rs373073146 Chr.5: 76954311-76954311 on Build GRCh38
rs1048498020 Chr.5: 76954321-76954321 on Build GRCh38
rs748509980 Chr.5: 76954229-76954229 on Build GRCh38
rs911261543 Chr.5: 76954337-76954337 on Build GRCh38
rs761765074 Chr.5: 76954171-76954171 on Build GRCh38
rs960533412 Chr.5: 76954263-76954263 on Build GRCh38
rs1020640444 Chr.5: 76954289-76954289 on Build GRCh38
rs761806124 Chr.5: 76954224-76954224 on Build GRCh38
rs574077513 Chr.5: 76954280-76954280 on Build GRCh38
rs377735644 Chr.5: 76954327-76954327 on Build GRCh38

View Full Product Details