Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60152948-60153213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7
Entrez Gene ID
51635
Gene Name
dehydrogenase/reductase 7
Gene Aliases
CGI-86, SDR34C1, retDSR4, retSDR4
UniGene
Hs.59719
Gene Chromosome Location
Chr.14: 60144119-60169864 on Build GRCh38
Transcripts
NM_001322280.1 NM_001322281.1 NM_001322282.1 NM_016029.3 XM_017021368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3815017 c.526C>T p.H176Y Chr.14: 60153046-60153046 on Build GRCh38
7140247 c.520C>A p.L174M Chr.14: 60153052-60153052 on Build GRCh38
6847237 c.545A>C p.Q182P Chr.14: 60153027-60153027 on Build GRCh38
5913566 c.514T>G p.C172G Chr.14: 60153058-60153058 on Build GRCh38
1323466 c.425C>T p.S142F Chr.14: 60153147-60153147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779784047 Chr.14: 60153075-60153075 on Build GRCh38
rs763765809 Chr.14: 60153134-60153134 on Build GRCh38
rs765181578 Chr.14: 60153148-60153148 on Build GRCh38
rs759548011 Chr.14: 60153103-60153103 on Build GRCh38
rs756537246 Chr.14: 60153177-60153177 on Build GRCh38
rs147318951 Chr.14: 60153159-60153159 on Build GRCh38
rs779337723 Chr.14: 60153190-60153190 on Build GRCh38
rs150536990 Chr.14: 60152985-60152985 on Build GRCh38
rs375076028 Chr.14: 60152995-60152995 on Build GRCh38
rs367690213 Chr.14: 60153099-60153099 on Build GRCh38
rs140180368 Chr.14: 60153074-60153074 on Build GRCh38
rs566717165 Chr.14: 60153187-60153187 on Build GRCh38
rs761678548 Chr.14: 60153146-60153146 on Build GRCh38
rs748482262 Chr.14: 60153017-60153017 on Build GRCh38
rs949059232 Chr.14: 60152992-60152992 on Build GRCh38
rs745485620 Chr.14: 60152989-60152989 on Build GRCh38
rs772297539 Chr.14: 60153108-60153108 on Build GRCh38
rs1033206846 Chr.14: 60153023-60153023 on Build GRCh38
rs199760290 Chr.14: 60152981-60152981 on Build GRCh38
rs142725427 Chr.14: 60153036-60153036 on Build GRCh38
rs375896675 Chr.14: 60153153-60153153 on Build GRCh38
rs770865940 Chr.14: 60153157-60153157 on Build GRCh38
rs190571329 Chr.14: 60153085-60153085 on Build GRCh38
rs371472289 Chr.14: 60153133-60153133 on Build GRCh38
rs146061320 Chr.14: 60153037-60153037 on Build GRCh38
rs773531485 Chr.14: 60153018-60153018 on Build GRCh38
rs772037175 Chr.14: 60153185-60153189 on Build GRCh38
rs138251248 Chr.14: 60153024-60153024 on Build GRCh38
rs111656127 Chr.14: 60153082-60153082 on Build GRCh38
rs1052931725 Chr.14: 60153130-60153130 on Build GRCh38
rs539643284 Chr.14: 60153141-60153141 on Build GRCh38
rs368159355 Chr.14: 60153035-60153035 on Build GRCh38
rs771423612 Chr.14: 60153096-60153096 on Build GRCh38
rs778406596 Chr.14: 60153087-60153087 on Build GRCh38
rs148567335 Chr.14: 60153149-60153149 on Build GRCh38
rs753717156 Chr.14: 60153043-60153043 on Build GRCh38
rs769191663 Chr.14: 60152994-60152994 on Build GRCh38
rs766188972 Chr.14: 60153175-60153175 on Build GRCh38
rs760406577 Chr.14: 60153042-60153042 on Build GRCh38
rs767916906 Chr.14: 60153136-60153136 on Build GRCh38
rs756832644 Chr.14: 60153046-60153046 on Build GRCh38
rs559221999 Chr.14: 60153038-60153038 on Build GRCh38
rs760586938 Chr.14: 60153129-60153129 on Build GRCh38
rs372021094 Chr.14: 60153052-60153052 on Build GRCh38
rs776680689 Chr.14: 60153142-60153142 on Build GRCh38
rs61742277 Chr.14: 60153092-60153092 on Build GRCh38
rs919468023 Chr.14: 60152996-60152996 on Build GRCh38
rs151158583 Chr.14: 60153176-60153176 on Build GRCh38
rs749902791 Chr.14: 60153055-60153055 on Build GRCh38
rs145805689 Chr.14: 60153119-60153119 on Build GRCh38
rs748533483 Chr.14: 60153076-60153076 on Build GRCh38
rs749627593 Chr.14: 60153186-60153186 on Build GRCh38
rs777974050 Chr.14: 60153178-60153178 on Build GRCh38
rs3180983 Chr.14: 60153116-60153116 on Build GRCh38

View Full Product Details