Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93871961-93872190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNTTIP2
Entrez Gene ID
30836
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Gene Aliases
ERBP, FCF2, HSU15552, LPTS-RP2, TdIF2
UniGene
Hs.601998
Gene Chromosome Location
Chr.1: 93869458-93879206 on Build GRCh38
Transcripts
NM_014597.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4634424 c.1953A>G p.P651P Chr.1: 93872186-93872186 on Build GRCh38
3493439 c.2014C>T p.P672S Chr.1: 93872125-93872125 on Build GRCh38
7147289 c.1971G>C p.L657L Chr.1: 93872168-93872168 on Build GRCh38
3493440 c.1969C>T p.L657L Chr.1: 93872170-93872170 on Build GRCh38
913423 c.2016G>T p.P672P Chr.1: 93872123-93872123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931437488 Chr.1: 93872138-93872138 on Build GRCh38
rs757456167 Chr.1: 93872103-93872103 on Build GRCh38
rs768025681 Chr.1: 93872131-93872131 on Build GRCh38
rs535246915 Chr.1: 93872054-93872054 on Build GRCh38
rs772951259 Chr.1: 93872046-93872046 on Build GRCh38
rs370834042 Chr.1: 93872057-93872057 on Build GRCh38
rs780700023 Chr.1: 93872126-93872126 on Build GRCh38
rs367746927 Chr.1: 93872124-93872124 on Build GRCh38
rs202045634 Chr.1: 93872082-93872082 on Build GRCh38
rs867514282 Chr.1: 93872041-93872041 on Build GRCh38
rs771784966 Chr.1: 93872039-93872039 on Build GRCh38
rs199741695 Chr.1: 93872024-93872024 on Build GRCh38
rs181128379 Chr.1: 93872048-93872048 on Build GRCh38
rs375350978 Chr.1: 93872064-93872064 on Build GRCh38
rs868395569 Chr.1: 93872158-93872158 on Build GRCh38
rs200778406 Chr.1: 93872146-93872146 on Build GRCh38
rs979966900 Chr.1: 93872108-93872108 on Build GRCh38
rs201567136 Chr.1: 93872130-93872130 on Build GRCh38
rs12748154 Chr.1: 93872112-93872112 on Build GRCh38
rs778437106 Chr.1: 93872144-93872144 on Build GRCh38
rs373677821 Chr.1: 93872123-93872123 on Build GRCh38

View Full Product Details