Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101608750-101609023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPCD
Entrez Gene ID
25911
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
Gene Aliases
-
UniGene
Hs.658128
Gene Chromosome Location
Chr.10: 101588332-101609653 on Build GRCh38
Transcripts
NM_015448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3806316 c.507+10A>C p.? Chr.10: 101608947-101608947 on Build GRCh38
4853505 c.501C>T p.I167I Chr.10: 101608931-101608931 on Build GRCh38
3751493 c.467T>C p.L156S Chr.10: 101608897-101608897 on Build GRCh38
5826589 c.508T>C p.C170R Chr.10: 101608897-101608897 on Build GRCh38
4844476 c.471G>A p.L157L Chr.10: 101608901-101608901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760598308 Chr.10: 101608832-101608832 on Build GRCh38
rs753260026 Chr.10: 101608885-101608885 on Build GRCh38
rs947747466 Chr.10: 101608952-101608952 on Build GRCh38
rs879813339 Chr.10: 101609001-101609001 on Build GRCh38
rs778449541 Chr.10: 101608799-101608799 on Build GRCh38
rs771096591 Chr.10: 101608835-101608835 on Build GRCh38
rs988166082 Chr.10: 101608868-101608868 on Build GRCh38
rs141479514 Chr.10: 101608938-101608938 on Build GRCh38
rs776550996 Chr.10: 101608839-101608839 on Build GRCh38
rs764611749 Chr.10: 101608947-101608947 on Build GRCh38
rs1018588385 Chr.10: 101608775-101608775 on Build GRCh38
rs752916349 Chr.10: 101608859-101608859 on Build GRCh38
rs371246163 Chr.10: 101608778-101608778 on Build GRCh38
rs1047243433 Chr.10: 101608976-101608976 on Build GRCh38
rs373259366 Chr.10: 101608866-101608866 on Build GRCh38
rs754448609 Chr.10: 101608970-101608970 on Build GRCh38
rs778723998 Chr.10: 101608814-101608814 on Build GRCh38
rs773135428 Chr.10: 101608823-101608823 on Build GRCh38
rs759842555 Chr.10: 101608842-101608842 on Build GRCh38
rs765408100 Chr.10: 101608847-101608847 on Build GRCh38
rs542483454 Chr.10: 101608892-101608892 on Build GRCh38
rs7006 Chr.10: 101608897-101608897 on Build GRCh38
rs757691660 Chr.10: 101608869-101608869 on Build GRCh38
rs61874047 Chr.10: 101608904-101608904 on Build GRCh38
rs373385453 Chr.10: 101608922-101608922 on Build GRCh38
rs371228299 Chr.10: 101608807-101608807 on Build GRCh38
rs377444359 Chr.10: 101608965-101608965 on Build GRCh38
rs769855917 Chr.10: 101608926-101608926 on Build GRCh38
rs775861439 Chr.10: 101608936-101608936 on Build GRCh38
rs957477615 Chr.10: 101608828-101608828 on Build GRCh38
rs754808265 Chr.10: 101608937-101608938 on Build GRCh38
rs375932631 Chr.10: 101608802-101608802 on Build GRCh38
rs558583763 Chr.10: 101608949-101608949 on Build GRCh38
rs778520556 Chr.10: 101608954-101608956 on Build GRCh38
rs764719218 Chr.10: 101608984-101608984 on Build GRCh38
rs201500851 Chr.10: 101608900-101608900 on Build GRCh38
rs747971984 Chr.10: 101608821-101608821 on Build GRCh38
rs140479530 Chr.10: 101608863-101608863 on Build GRCh38
rs928651278 Chr.10: 101608910-101608910 on Build GRCh38
rs369112227 Chr.10: 101608822-101608822 on Build GRCh38
rs145149344 Chr.10: 101608913-101608913 on Build GRCh38
rs758252691 Chr.10: 101608898-101608898 on Build GRCh38
rs763426295 Chr.10: 101608860-101608860 on Build GRCh38
rs35578063 Chr.10: 101608972-101608973 on Build GRCh38
rs779426654 Chr.10: 101608806-101608806 on Build GRCh38
rs752180220 Chr.10: 101608864-101608864 on Build GRCh38
rs374959612 Chr.10: 101608812-101608812 on Build GRCh38
rs776590195 Chr.10: 101608918-101608918 on Build GRCh38
rs200230925 Chr.10: 101608840-101608840 on Build GRCh38
rs752209043 Chr.10: 101608975-101608976 on Build GRCh38
rs371576475 Chr.10: 101608963-101608963 on Build GRCh38
rs762425987 Chr.10: 101608960-101608960 on Build GRCh38
rs577251111 Chr.10: 101608961-101608961 on Build GRCh38
rs779668087 Chr.10: 101608873-101608873 on Build GRCh38
rs375546319 Chr.10: 101608893-101608893 on Build GRCh38
rs773833827 Chr.10: 101608914-101608914 on Build GRCh38
rs1005370477 Chr.10: 101608801-101608801 on Build GRCh38
rs758794132 Chr.10: 101608784-101608784 on Build GRCh38

View Full Product Details