Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142595261-142595530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF1
Entrez Gene ID
2246
Gene Name
fibroblast growth factor 1
Gene Aliases
AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1
UniGene
Hs.483635
Gene Chromosome Location
Chr.5: 142592178-142698070 on Build GRCh38
Transcripts
NM_000800.4 NM_001144892.2 NM_001144934.1 NM_001144935.1 NM_001257205.1 NM_001257206.1 NM_001257207.1 NM_001257208.1 NM_001257209.1 NM_001257210.1 NM_001257211.1 NM_001257212.1 NM_033136.3 NM_033137.2 XM_005268390.4 XM_011537607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3994079 c.454G>T p.V152F Chr.5: 142595304-142595304 on Build GRCh38
1207046 c.403G>A p.G135S Chr.5: 142595355-142595355 on Build GRCh38
4495384 c.452C>T p.P151L Chr.5: 142595306-142595306 on Build GRCh38
6663460 c.400C>T p.R134C Chr.5: 142595358-142595358 on Build GRCh38
1434817 c.329A>C p.N110T Chr.5: 142595429-142595429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989785171 Chr.5: 142595501-142595501 on Build GRCh38
rs748034939 Chr.5: 142595324-142595324 on Build GRCh38
rs780390744 Chr.5: 142595392-142595392 on Build GRCh38
rs769092526 Chr.5: 142595470-142595470 on Build GRCh38
rs753037736 Chr.5: 142595310-142595310 on Build GRCh38
rs765790331 Chr.5: 142595493-142595493 on Build GRCh38
rs754699487 Chr.5: 142595319-142595319 on Build GRCh38
rs754388820 Chr.5: 142595361-142595361 on Build GRCh38
rs770590235 Chr.5: 142595349-142595349 on Build GRCh38
rs554308959 Chr.5: 142595308-142595308 on Build GRCh38
rs757372975 Chr.5: 142595312-142595312 on Build GRCh38
rs369740938 Chr.5: 142595406-142595406 on Build GRCh38
rs373883111 Chr.5: 142595458-142595458 on Build GRCh38
rs746120174 Chr.5: 142595317-142595317 on Build GRCh38
rs34003 Chr.5: 142595502-142595502 on Build GRCh38
rs769616526 Chr.5: 142595325-142595325 on Build GRCh38
rs777523983 Chr.5: 142595422-142595422 on Build GRCh38
rs923410105 Chr.5: 142595288-142595288 on Build GRCh38
rs376396733 Chr.5: 142595389-142595389 on Build GRCh38
rs761839384 Chr.5: 142595364-142595364 on Build GRCh38
rs1043307908 Chr.5: 142595380-142595380 on Build GRCh38
rs17223954 Chr.5: 142595487-142595487 on Build GRCh38
rs141473407 Chr.5: 142595305-142595305 on Build GRCh38
rs201277854 Chr.5: 142595315-142595315 on Build GRCh38
rs750586330 Chr.5: 142595368-142595368 on Build GRCh38
rs770501980 Chr.5: 142595436-142595436 on Build GRCh38
rs780860378 Chr.5: 142595320-142595320 on Build GRCh38
rs146745580 Chr.5: 142595348-142595348 on Build GRCh38
rs759270544 Chr.5: 142595355-142595355 on Build GRCh38
rs745432170 Chr.5: 142595455-142595455 on Build GRCh38
rs145843967 Chr.5: 142595357-142595357 on Build GRCh38
rs143011306 Chr.5: 142595463-142595463 on Build GRCh38
rs143888146 Chr.5: 142595341-142595341 on Build GRCh38
rs764484123 Chr.5: 142595358-142595358 on Build GRCh38
rs149033545 Chr.5: 142595425-142595425 on Build GRCh38
rs372965702 Chr.5: 142595356-142595356 on Build GRCh38
rs775253006 Chr.5: 142595461-142595461 on Build GRCh38
rs765303323 Chr.5: 142595366-142595366 on Build GRCh38
rs902995255 Chr.5: 142595384-142595384 on Build GRCh38
rs561960626 Chr.5: 142595421-142595421 on Build GRCh38
rs764725960 Chr.5: 142595310-142595310 on Build GRCh38
rs778584449 Chr.5: 142595449-142595449 on Build GRCh38
rs774109079 Chr.5: 142595352-142595352 on Build GRCh38
rs777053892 Chr.5: 142595486-142595486 on Build GRCh38

View Full Product Details