Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86565733-86565966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA4
Entrez Gene ID
22802
Gene Name
chloride channel accessory 4
Gene Aliases
CaCC, CaCC2
UniGene
Hs.567422
Gene Chromosome Location
Chr.1: 86547076-86580758 on Build GRCh38
Transcripts
NM_012128.3 NR_024602.1 XM_011541015.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6127960 c.23C>T p.P8L Chr.1: 86565950-86565950 on Build GRCh38
6064616 c.784C>A p.L262I Chr.1: 86565850-86565850 on Build GRCh38
6305840 c.6G>T p.V2V Chr.1: 86565933-86565933 on Build GRCh38
5702991 c.739G>A p.V247I Chr.1: 86565805-86565805 on Build GRCh38
2244482 c.754G>A p.E252K Chr.1: 86565820-86565820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77380561 Chr.1: 86565895-86565895 on Build GRCh38
rs545676551 Chr.1: 86565805-86565805 on Build GRCh38
rs200731167 Chr.1: 86565766-86565766 on Build GRCh38
rs952121137 Chr.1: 86565925-86565925 on Build GRCh38
rs755357644 Chr.1: 86565867-86565867 on Build GRCh38
rs377545104 Chr.1: 86565904-86565904 on Build GRCh38
rs754154848 Chr.1: 86565866-86565866 on Build GRCh38
rs760725513 Chr.1: 86565860-86565860 on Build GRCh38
rs992171986 Chr.1: 86565913-86565913 on Build GRCh38
rs962354275 Chr.1: 86565910-86565910 on Build GRCh38
rs766688618 Chr.1: 86565864-86565864 on Build GRCh38
rs745628367 Chr.1: 86565802-86565802 on Build GRCh38
rs779532185 Chr.1: 86565768-86565768 on Build GRCh38
rs775266586 Chr.1: 86565928-86565928 on Build GRCh38
rs186153841 Chr.1: 86565886-86565886 on Build GRCh38
rs922057500 Chr.1: 86565897-86565897 on Build GRCh38
rs748724090 Chr.1: 86565769-86565769 on Build GRCh38
rs1018750206 Chr.1: 86565828-86565828 on Build GRCh38
rs965913722 Chr.1: 86565861-86565861 on Build GRCh38
rs756797338 Chr.1: 86565923-86565923 on Build GRCh38
rs757799842 Chr.1: 86565778-86565778 on Build GRCh38
rs776312652 Chr.1: 86565798-86565798 on Build GRCh38
rs765721412 Chr.1: 86565882-86565882 on Build GRCh38
rs746501996 Chr.1: 86565920-86565920 on Build GRCh38
rs775355339 Chr.1: 86565807-86565807 on Build GRCh38
rs373050506 Chr.1: 86565842-86565842 on Build GRCh38
rs781062541 Chr.1: 86565820-86565821 on Build GRCh38
rs753092233 Chr.1: 86565889-86565889 on Build GRCh38
rs765220655 Chr.1: 86565827-86565827 on Build GRCh38
rs754244572 Chr.1: 86565764-86565764 on Build GRCh38
rs916137005 Chr.1: 86565916-86565916 on Build GRCh38
rs955974655 Chr.1: 86565801-86565801 on Build GRCh38
rs746564354 Chr.1: 86565792-86565792 on Build GRCh38
rs199899112 Chr.1: 86565819-86565819 on Build GRCh38
rs758951749 Chr.1: 86565896-86565896 on Build GRCh38
rs777359179 Chr.1: 86565788-86565788 on Build GRCh38
rs908124082 Chr.1: 86565941-86565941 on Build GRCh38
rs971981455 Chr.1: 86565929-86565929 on Build GRCh38
rs770599952 Chr.1: 86565794-86565794 on Build GRCh38
rs780779596 Chr.1: 86565926-86565926 on Build GRCh38
rs951147671 Chr.1: 86565835-86565835 on Build GRCh38

View Full Product Details