Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 154410312-154410537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR3
Entrez Gene ID
154043
Gene Name
CNKSR family member 3
Gene Aliases
MAGI1
UniGene
Hs.16064
Gene Chromosome Location
Chr.6: 154405299-154510619 on Build GRCh38
Transcripts
NM_173515.2 XM_011535485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1248557 c.1361G>A p.G454D Chr.6: 154410351-154410351 on Build GRCh38
4807043 c.1312G>A p.G438R Chr.6: 154410400-154410400 on Build GRCh38
305615 c.1284G>A p.K428K Chr.6: 154410428-154410428 on Build GRCh38
1621119 c.1283A>G p.K428R Chr.6: 154410429-154410429 on Build GRCh38
6631254 c.1341delT p.F447fs*107 Chr.6: 154410371-154410371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201171745 Chr.6: 154410445-154410445 on Build GRCh38
rs766371759 Chr.6: 154410352-154410352 on Build GRCh38
rs199966980 Chr.6: 154410444-154410444 on Build GRCh38
rs895190263 Chr.6: 154410391-154410391 on Build GRCh38
rs186911920 Chr.6: 154410432-154410432 on Build GRCh38
rs988599292 Chr.6: 154410511-154410511 on Build GRCh38
rs964643669 Chr.6: 154410485-154410485 on Build GRCh38
rs374933467 Chr.6: 154410414-154410414 on Build GRCh38
rs777324452 Chr.6: 154410412-154410412 on Build GRCh38
rs772775895 Chr.6: 154410442-154410442 on Build GRCh38
rs747147689 Chr.6: 154410399-154410399 on Build GRCh38
rs754818245 Chr.6: 154410358-154410358 on Build GRCh38
rs974538894 Chr.6: 154410491-154410491 on Build GRCh38
rs777910802 Chr.6: 154410477-154410477 on Build GRCh38
rs765091975 Chr.6: 154410463-154410463 on Build GRCh38
rs753619526 Chr.6: 154410357-154410357 on Build GRCh38
rs750273939 Chr.6: 154410348-154410348 on Build GRCh38
rs771548567 Chr.6: 154410404-154410404 on Build GRCh38
rs200910224 Chr.6: 154410339-154410339 on Build GRCh38
rs557511874 Chr.6: 154410337-154410337 on Build GRCh38
rs756049782 Chr.6: 154410349-154410349 on Build GRCh38
rs139168826 Chr.6: 154410369-154410369 on Build GRCh38
rs954609191 Chr.6: 154410508-154410508 on Build GRCh38
rs753815146 Chr.6: 154410454-154410454 on Build GRCh38
rs747059735 Chr.6: 154410482-154410482 on Build GRCh38
rs777913554 Chr.6: 154410385-154410385 on Build GRCh38
rs770316254 Chr.6: 154410416-154410416 on Build GRCh38
rs149933492 Chr.6: 154410423-154410423 on Build GRCh38
rs542942542 Chr.6: 154410438-154410438 on Build GRCh38
rs1045676575 Chr.6: 154410347-154410347 on Build GRCh38
rs543339316 Chr.6: 154410360-154410360 on Build GRCh38
rs748423356 Chr.6: 154410367-154410367 on Build GRCh38
rs751600510 Chr.6: 154410499-154410499 on Build GRCh38
rs4502943 Chr.6: 154410513-154410513 on Build GRCh38
rs751267724 Chr.6: 154410341-154410341 on Build GRCh38
rs369633632 Chr.6: 154410338-154410338 on Build GRCh38
rs78244842 Chr.6: 154410346-154410346 on Build GRCh38
rs761192211 Chr.6: 154410344-154410344 on Build GRCh38
rs540745131 Chr.6: 154410464-154410464 on Build GRCh38
rs754717721 Chr.6: 154410461-154410461 on Build GRCh38

View Full Product Details