Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59152904-59153169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM111A
Entrez Gene ID
63901
Gene Name
family with sequence similarity 111 member A
Gene Aliases
GCLEB, KCS2
UniGene
Hs.150651
Gene Chromosome Location
Chr.11: 59142746-59155039 on Build GRCh38
Transcripts
NM_001142519.2 NM_001142520.2 NM_001142521.2 NM_001312909.1 NM_001312910.1 NM_001312911.1 NM_022074.3 NM_198847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7165358 c.1410T>C p.H470H Chr.11: 59153078-59153078 on Build GRCh38
6658088 c.1476G>A p.Q492Q Chr.11: 59153144-59153144 on Build GRCh38
6588120 c.1461G>A p.V487V Chr.11: 59153129-59153129 on Build GRCh38
1475589 c.1481C>G p.A494G Chr.11: 59153149-59153149 on Build GRCh38
5901712 c.1286C>G p.P429R Chr.11: 59152954-59152954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199632362 Chr.11: 59153130-59153130 on Build GRCh38
rs113832524 Chr.11: 59152991-59152991 on Build GRCh38
rs763271241 Chr.11: 59153018-59153019 on Build GRCh38
rs762523325 Chr.11: 59153014-59153014 on Build GRCh38
rs147361273 Chr.11: 59153076-59153076 on Build GRCh38
rs964003230 Chr.11: 59153108-59153108 on Build GRCh38
rs768479872 Chr.11: 59153080-59153080 on Build GRCh38
rs774796498 Chr.11: 59153018-59153018 on Build GRCh38
rs565765996 Chr.11: 59153127-59153127 on Build GRCh38
rs746124549 Chr.11: 59153053-59153053 on Build GRCh38
rs978791629 Chr.11: 59153083-59153083 on Build GRCh38
rs375431912 Chr.11: 59153113-59153113 on Build GRCh38
rs769752882 Chr.11: 59153110-59153110 on Build GRCh38
rs372448906 Chr.11: 59153050-59153050 on Build GRCh38
rs960606232 Chr.11: 59152993-59152993 on Build GRCh38
rs116918730 Chr.11: 59153019-59153019 on Build GRCh38
rs764535228 Chr.11: 59153146-59153146 on Build GRCh38
rs757426077 Chr.11: 59153029-59153029 on Build GRCh38
rs760805581 Chr.11: 59153134-59153134 on Build GRCh38
rs1018329469 Chr.11: 59153106-59153106 on Build GRCh38
rs749524786 Chr.11: 59152936-59152936 on Build GRCh38
rs761459675 Chr.11: 59152989-59152989 on Build GRCh38
rs761191576 Chr.11: 59153145-59153145 on Build GRCh38
rs149508004 Chr.11: 59152953-59152953 on Build GRCh38
rs764994003 Chr.11: 59152997-59152997 on Build GRCh38
rs767475718 Chr.11: 59153102-59153102 on Build GRCh38
rs772115241 Chr.11: 59153061-59153061 on Build GRCh38
rs749914836 Chr.11: 59153003-59153003 on Build GRCh38
rs754170888 Chr.11: 59153028-59153028 on Build GRCh38
rs12224380 Chr.11: 59153144-59153144 on Build GRCh38
rs368508077 Chr.11: 59152955-59152955 on Build GRCh38
rs148396354 Chr.11: 59153133-59153133 on Build GRCh38
rs142586710 Chr.11: 59153135-59153135 on Build GRCh38
rs114634830 Chr.11: 59152967-59152967 on Build GRCh38
rs771954122 Chr.11: 59152954-59152954 on Build GRCh38
rs780052713 Chr.11: 59153063-59153063 on Build GRCh38
rs764485455 Chr.11: 59153019-59153020 on Build GRCh38
rs748806972 Chr.11: 59153088-59153088 on Build GRCh38
rs1035926722 Chr.11: 59152987-59152987 on Build GRCh38
rs147219534 Chr.11: 59152964-59152964 on Build GRCh38
rs771148654 Chr.11: 59152940-59152940 on Build GRCh38
rs778134736 Chr.11: 59153022-59153022 on Build GRCh38
rs776357022 Chr.11: 59152969-59152969 on Build GRCh38
rs761722101 Chr.11: 59153109-59153109 on Build GRCh38

View Full Product Details