Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24548756-24549029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCD1
Entrez Gene ID
83606
Gene Name
guanylyl cyclase domain containing 1
Gene Aliases
C22orf13, LLN4
UniGene
Hs.9850
Gene Chromosome Location
Chr.22: 24540423-24555935 on Build GRCh38
Transcripts
NM_001284251.1 NM_001284252.1 NM_001284253.1 NM_001284254.1 NM_001284255.1 NM_001284256.1 NM_001284257.1 NM_031444.3 NR_104286.1 XM_005261761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6566092 c.90C>T p.H30H Chr.22: 24548955-24548955 on Build GRCh38
6253574 c.127C>A p.R43R Chr.22: 24548918-24548918 on Build GRCh38
1032728 c.69C>T p.P23P Chr.22: 24548976-24548976 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765456298 Chr.22: 24548819-24548819 on Build GRCh38
rs867148079 Chr.22: 24548999-24548999 on Build GRCh38
rs181521384 Chr.22: 24548845-24548845 on Build GRCh38
rs747594368 Chr.22: 24548976-24548976 on Build GRCh38
rs200818390 Chr.22: 24548959-24548959 on Build GRCh38
rs1042599549 Chr.22: 24548829-24548829 on Build GRCh38
rs759509576 Chr.22: 24548886-24548886 on Build GRCh38
rs371487432 Chr.22: 24548979-24548979 on Build GRCh38
rs556283469 Chr.22: 24548910-24548910 on Build GRCh38
rs756449863 Chr.22: 24548918-24548918 on Build GRCh38
rs544188933 Chr.22: 24548902-24548902 on Build GRCh38
rs1050187874 Chr.22: 24548904-24548904 on Build GRCh38
rs367688728 Chr.22: 24548897-24548897 on Build GRCh38
rs970405969 Chr.22: 24548882-24548882 on Build GRCh38
rs754372061 Chr.22: 24548943-24548943 on Build GRCh38
rs141689012 Chr.22: 24548975-24548975 on Build GRCh38
rs1023967391 Chr.22: 24548870-24548870 on Build GRCh38
rs756257993 Chr.22: 24548898-24548899 on Build GRCh38
rs16978648 Chr.22: 24548782-24548782 on Build GRCh38
rs780346021 Chr.22: 24548931-24548931 on Build GRCh38
rs925793920 Chr.22: 24548972-24548972 on Build GRCh38
rs764313417 Chr.22: 24548889-24548889 on Build GRCh38
rs1003580 Chr.22: 24548814-24548814 on Build GRCh38
rs553914429 Chr.22: 24548917-24548917 on Build GRCh38
rs995007458 Chr.22: 24548788-24548788 on Build GRCh38

View Full Product Details