Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10994954-10995227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL2
Entrez Gene ID
54898
Gene Name
ELOVL fatty acid elongase 2
Gene Aliases
SSC2
UniGene
Hs.656436
Gene Chromosome Location
Chr.6: 10980759-11044391 on Build GRCh38
Transcripts
NM_017770.3 XM_011514716.2 XM_011514717.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3155550 c.479T>C p.V160A Chr.6: 10995033-10995033 on Build GRCh38
4903142 c.396C>T p.F132F Chr.6: 10995116-10995116 on Build GRCh38
4916408 c.391A>C p.I131L Chr.6: 10995121-10995121 on Build GRCh38
3155552 c.373G>A p.V125I Chr.6: 10995139-10995139 on Build GRCh38
5404149 c.500G>A p.G167E Chr.6: 10995012-10995012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747947657 Chr.6: 10995199-10995199 on Build GRCh38
rs771927185 Chr.6: 10995200-10995200 on Build GRCh38
rs774368530 Chr.6: 10995001-10995001 on Build GRCh38
rs761880960 Chr.6: 10995107-10995107 on Build GRCh38
rs542625138 Chr.6: 10995118-10995118 on Build GRCh38
rs1029567955 Chr.6: 10995149-10995149 on Build GRCh38
rs765604632 Chr.6: 10995127-10995127 on Build GRCh38
rs369583586 Chr.6: 10995002-10995002 on Build GRCh38
rs768479188 Chr.6: 10995088-10995088 on Build GRCh38
rs765612044 Chr.6: 10995041-10995041 on Build GRCh38
rs749968212 Chr.6: 10995060-10995060 on Build GRCh38
rs141887901 Chr.6: 10995095-10995095 on Build GRCh38
rs754080552 Chr.6: 10995025-10995025 on Build GRCh38
rs377540194 Chr.6: 10995074-10995074 on Build GRCh38
rs753501622 Chr.6: 10995178-10995178 on Build GRCh38
rs1003874102 Chr.6: 10995181-10995181 on Build GRCh38
rs759724801 Chr.6: 10995122-10995122 on Build GRCh38
rs780837408 Chr.6: 10995058-10995058 on Build GRCh38
rs971114052 Chr.6: 10995062-10995062 on Build GRCh38
rs778753765 Chr.6: 10995189-10995189 on Build GRCh38
rs373748893 Chr.6: 10995004-10995004 on Build GRCh38
rs867205847 Chr.6: 10995027-10995027 on Build GRCh38
rs755711524 Chr.6: 10995153-10995153 on Build GRCh38
rs955064791 Chr.6: 10995171-10995171 on Build GRCh38
rs753223918 Chr.6: 10995044-10995044 on Build GRCh38
rs749117263 Chr.6: 10994988-10994988 on Build GRCh38
rs376129618 Chr.6: 10994983-10994983 on Build GRCh38
rs775370968 Chr.6: 10994987-10994987 on Build GRCh38
rs776914003 Chr.6: 10995036-10995036 on Build GRCh38
rs777715421 Chr.6: 10995202-10995202 on Build GRCh38
rs760870471 Chr.6: 10995015-10995015 on Build GRCh38
rs773344524 Chr.6: 10995007-10995007 on Build GRCh38
rs768438687 Chr.6: 10994994-10994994 on Build GRCh38
rs757950376 Chr.6: 10995007-10995007 on Build GRCh38
rs147119821 Chr.6: 10995170-10995170 on Build GRCh38
rs780896280 Chr.6: 10994982-10994982 on Build GRCh38
rs755762314 Chr.6: 10995066-10995066 on Build GRCh38
rs867506828 Chr.6: 10995106-10995106 on Build GRCh38
rs766597383 Chr.6: 10995024-10995024 on Build GRCh38
rs777338409 Chr.6: 10995089-10995090 on Build GRCh38
rs772052269 Chr.6: 10995097-10995097 on Build GRCh38
rs769883226 Chr.6: 10995121-10995121 on Build GRCh38
rs147952046 Chr.6: 10995071-10995071 on Build GRCh38
rs374486290 Chr.6: 10995113-10995113 on Build GRCh38
rs763497968 Chr.6: 10995137-10995137 on Build GRCh38
rs572165008 Chr.6: 10995105-10995105 on Build GRCh38
rs367977328 Chr.6: 10995182-10995182 on Build GRCh38
rs371048408 Chr.6: 10995079-10995079 on Build GRCh38
rs756794823 Chr.6: 10995113-10995115 on Build GRCh38
rs748990852 Chr.6: 10995083-10995083 on Build GRCh38
rs749913053 Chr.6: 10995146-10995146 on Build GRCh38
rs751245099 Chr.6: 10995096-10995097 on Build GRCh38
rs755401151 Chr.6: 10995031-10995031 on Build GRCh38
rs553897838 Chr.6: 10995050-10995050 on Build GRCh38
rs771020800 Chr.6: 10995207-10995207 on Build GRCh38
rs893190741 Chr.6: 10995034-10995034 on Build GRCh38
rs996804777 Chr.6: 10995109-10995109 on Build GRCh38
rs202213512 Chr.6: 10994984-10994984 on Build GRCh38
rs753067950 Chr.6: 10995132-10995132 on Build GRCh38
rs764703919 Chr.6: 10995138-10995138 on Build GRCh38
rs748114809 Chr.6: 10995096-10995096 on Build GRCh38

View Full Product Details