Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 101178038-101178263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHSY1
Entrez Gene ID
22856
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
CHSY, CSS1, ChSy-1, TPBS
UniGene
Hs.110488
Gene Chromosome Location
Chr.15: 101175723-101251932 on Build GRCh38
Transcripts
NM_014918.4 XM_006720435.3 XM_011521364.2 XM_017022011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7301666 c.1660T>C p.F554L Chr.15: 101178137-101178137 on Build GRCh38
5687466 c.1624G>T p.G542W Chr.15: 101178173-101178173 on Build GRCh38
3499764 c.1655G>A p.G552E Chr.15: 101178142-101178142 on Build GRCh38
3499763 c.1685A>G p.N562S Chr.15: 101178112-101178112 on Build GRCh38
7256046 c.1708C>T p.L570L Chr.15: 101178089-101178089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151237975 Chr.15: 101178072-101178072 on Build GRCh38
rs781661334 Chr.15: 101178104-101178104 on Build GRCh38
rs387906985 Chr.15: 101178181-101178181 on Build GRCh38
rs892627220 Chr.15: 101178113-101178113 on Build GRCh38
rs74784454 Chr.15: 101178231-101178231 on Build GRCh38
rs757907731 Chr.15: 101178060-101178060 on Build GRCh38
rs781478353 Chr.15: 101178129-101178129 on Build GRCh38
rs139276897 Chr.15: 101178080-101178080 on Build GRCh38
rs768173595 Chr.15: 101178170-101178170 on Build GRCh38
rs146586939 Chr.15: 101178195-101178195 on Build GRCh38
rs761401345 Chr.15: 101178108-101178108 on Build GRCh38
rs1041160406 Chr.15: 101178144-101178144 on Build GRCh38
rs192677621 Chr.15: 101178096-101178096 on Build GRCh38
rs767149155 Chr.15: 101178182-101178182 on Build GRCh38
rs767347072 Chr.15: 101178109-101178109 on Build GRCh38
rs762538148 Chr.15: 101178087-101178087 on Build GRCh38
rs141305214 Chr.15: 101178197-101178197 on Build GRCh38
rs548628862 Chr.15: 101178081-101178081 on Build GRCh38
rs552355921 Chr.15: 101178165-101178165 on Build GRCh38
rs776069033 Chr.15: 101178164-101178164 on Build GRCh38
rs769993716 Chr.15: 101178153-101178153 on Build GRCh38
rs754944859 Chr.15: 101178219-101178219 on Build GRCh38
rs766053226 Chr.15: 101178204-101178204 on Build GRCh38
rs1054078127 Chr.15: 101178099-101178099 on Build GRCh38
rs144268167 Chr.15: 101178126-101178126 on Build GRCh38
rs757609213 Chr.15: 101178240-101178240 on Build GRCh38
rs753655967 Chr.15: 101178117-101178117 on Build GRCh38
rs751688006 Chr.15: 101178232-101178232 on Build GRCh38
rs761491604 Chr.15: 101178176-101178176 on Build GRCh38
rs746229987 Chr.15: 101178146-101178146 on Build GRCh38
rs758585689 Chr.15: 101178122-101178122 on Build GRCh38
rs746282806 Chr.15: 101178112-101178112 on Build GRCh38
rs972865182 Chr.15: 101178178-101178178 on Build GRCh38
rs774020290 Chr.15: 101178100-101178100 on Build GRCh38
rs371244530 Chr.15: 101178188-101178188 on Build GRCh38
rs866476738 Chr.15: 101178095-101178095 on Build GRCh38
rs374805016 Chr.15: 101178208-101178208 on Build GRCh38
rs372414536 Chr.15: 101178079-101178079 on Build GRCh38
rs754947736 Chr.15: 101178242-101178242 on Build GRCh38
rs777115970 Chr.15: 101178074-101178074 on Build GRCh38
rs756067077 Chr.15: 101178116-101178116 on Build GRCh38
rs374869761 Chr.15: 101178127-101178127 on Build GRCh38
rs773824794 Chr.15: 101178175-101178175 on Build GRCh38

View Full Product Details