Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13029890-13030163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP192
Entrez Gene ID
55125
Gene Name
centrosomal protein 192
Gene Aliases
PPP1R62
UniGene
Hs.100914
Gene Chromosome Location
Chr.18: 12991307-13125053 on Build GRCh38
Transcripts
NM_032142.3 XM_005258107.3 XM_005258109.3 XM_005258110.1 XM_006722326.3 XM_006722327.3 XM_006722330.3 XM_011525673.2 XM_011525675.2 XM_017025803.1 XM_017025804.1 XM_017025805.1 XM_017025806.1 XM_017025807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6246968 c.1303G>T p.G435* Chr.18: 13029915-13029915 on Build GRCh38
5700751 c.1363C>T p.H455Y Chr.18: 13029975-13029975 on Build GRCh38
7449458 c.1310A>G p.N437S Chr.18: 13029922-13029922 on Build GRCh38
4597513 c.1375G>A p.E459K Chr.18: 13029987-13029987 on Build GRCh38
4151829 c.1335A>T p.P445P Chr.18: 13029947-13029947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756284733 Chr.18: 13030059-13030059 on Build GRCh38
rs887900150 Chr.18: 13029961-13029961 on Build GRCh38
rs919276850 Chr.18: 13029928-13029928 on Build GRCh38
rs116020861 Chr.18: 13029987-13029987 on Build GRCh38
rs763637873 Chr.18: 13030031-13030031 on Build GRCh38
rs767224438 Chr.18: 13030023-13030024 on Build GRCh38
rs556589449 Chr.18: 13030061-13030061 on Build GRCh38
rs909083098 Chr.18: 13030102-13030102 on Build GRCh38
rs527989767 Chr.18: 13029994-13029994 on Build GRCh38
rs1040621940 Chr.18: 13029925-13029925 on Build GRCh38
rs1031573843 Chr.18: 13030130-13030130 on Build GRCh38
rs776353014 Chr.18: 13029946-13029946 on Build GRCh38
rs765104857 Chr.18: 13029983-13029983 on Build GRCh38
rs756917482 Chr.18: 13030047-13030047 on Build GRCh38
rs758385798 Chr.18: 13030097-13030097 on Build GRCh38
rs781618337 Chr.18: 13030051-13030051 on Build GRCh38
rs552534235 Chr.18: 13029965-13029965 on Build GRCh38
rs79147242 Chr.18: 13030084-13030084 on Build GRCh38
rs750680465 Chr.18: 13030045-13030045 on Build GRCh38
rs543959589 Chr.18: 13030138-13030138 on Build GRCh38
rs535172505 Chr.18: 13030042-13030042 on Build GRCh38
rs1031385807 Chr.18: 13029945-13029945 on Build GRCh38
rs762414067 Chr.18: 13029989-13029989 on Build GRCh38
rs200779293 Chr.18: 13029948-13029948 on Build GRCh38

View Full Product Details