Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25207179-25207424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD6
Entrez Gene ID
955
Gene Name
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Gene Aliases
CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3
UniGene
Hs.500375
Gene Chromosome Location
Chr.20: 25195670-25228075 on Build GRCh38
Transcripts
NM_001114089.3 NM_001247.4 NM_001317941.2 NM_001322378.1 NM_001322379.1 NM_001322380.1 NM_001322381.1 NM_001322382.1 NM_001322383.1 NM_001322384.1 NM_001322385.1 NM_001322386.1 NM_001322387.1 NM_001322388.1 NM_001322389.1 NM_001322390.1 NM_001322391.1 NM_001322392.1 NM_001322393.1 NM_001322394.1 NM_001322395.1 NM_001322396.1 NM_001322397.1 NM_001322398.1 XM_006723665.2 XM_011529396.1 XM_011529397.1 XM_017028129.1 XM_017028130.1 XM_017028131.1 XM_017028132.1 XM_017028133.1 XM_017028134.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6092967 c.308G>T p.G103V Chr.20: 25207329-25207329 on Build GRCh38
4097254 c.275G>A p.G92E Chr.20: 25207296-25207296 on Build GRCh38
6092966 c.308G>T p.G103V Chr.20: 25207329-25207329 on Build GRCh38
4648378 c.288C>T p.D96D Chr.20: 25207309-25207309 on Build GRCh38
6159564 c.234C>G p.A78A Chr.20: 25207255-25207255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146711180 Chr.20: 25207257-25207257 on Build GRCh38
rs769657862 Chr.20: 25207387-25207388 on Build GRCh38
rs780239675 Chr.20: 25207382-25207382 on Build GRCh38
rs892359728 Chr.20: 25207252-25207252 on Build GRCh38
rs749957804 Chr.20: 25207231-25207231 on Build GRCh38
rs753301056 Chr.20: 25207404-25207404 on Build GRCh38
rs865899262 Chr.20: 25207260-25207260 on Build GRCh38
rs963683114 Chr.20: 25207240-25207240 on Build GRCh38
rs200024189 Chr.20: 25207220-25207220 on Build GRCh38
rs758678698 Chr.20: 25207326-25207326 on Build GRCh38
rs766990810 Chr.20: 25207229-25207229 on Build GRCh38
rs760258371 Chr.20: 25207400-25207400 on Build GRCh38
rs938534827 Chr.20: 25207272-25207272 on Build GRCh38
rs143602545 Chr.20: 25207265-25207265 on Build GRCh38
rs762648913 Chr.20: 25207286-25207286 on Build GRCh38
rs141279047 Chr.20: 25207212-25207212 on Build GRCh38
rs202026558 Chr.20: 25207328-25207328 on Build GRCh38
rs764004461 Chr.20: 25207288-25207288 on Build GRCh38
rs565323909 Chr.20: 25207219-25207219 on Build GRCh38
rs755787165 Chr.20: 25207239-25207239 on Build GRCh38
rs757746946 Chr.20: 25207336-25207336 on Build GRCh38
rs773322178 Chr.20: 25207397-25207397 on Build GRCh38
rs775741406 Chr.20: 25207282-25207282 on Build GRCh38
rs769926116 Chr.20: 25207378-25207378 on Build GRCh38
rs138746888 Chr.20: 25207386-25207386 on Build GRCh38
rs774025988 Chr.20: 25207292-25207292 on Build GRCh38
rs746473952 Chr.20: 25207251-25207251 on Build GRCh38
rs377433296 Chr.20: 25207250-25207250 on Build GRCh38
rs371107168 Chr.20: 25207289-25207289 on Build GRCh38
rs551145330 Chr.20: 25207211-25207211 on Build GRCh38
rs775434315 Chr.20: 25207388-25207388 on Build GRCh38
rs749465292 Chr.20: 25207384-25207384 on Build GRCh38
rs753612802 Chr.20: 25207320-25207320 on Build GRCh38
rs1044566 Chr.20: 25207234-25207234 on Build GRCh38
rs145732789 Chr.20: 25207370-25207370 on Build GRCh38
rs745640818 Chr.20: 25207323-25207324 on Build GRCh38
rs760309125 Chr.20: 25207309-25207309 on Build GRCh38
rs140692182 Chr.20: 25207296-25207296 on Build GRCh38
rs1045210152 Chr.20: 25207213-25207213 on Build GRCh38
rs771355768 Chr.20: 25207262-25207262 on Build GRCh38
rs767593042 Chr.20: 25207304-25207304 on Build GRCh38
rs766097746 Chr.20: 25207316-25207316 on Build GRCh38
rs766046799 Chr.20: 25207403-25207403 on Build GRCh38
rs749902985 Chr.20: 25207307-25207307 on Build GRCh38
rs35296506 Chr.20: 25207266-25207266 on Build GRCh38
rs374181395 Chr.20: 25207369-25207369 on Build GRCh38
rs1026467823 Chr.20: 25207390-25207390 on Build GRCh38
rs768761704 Chr.20: 25207385-25207385 on Build GRCh38
rs772110954 Chr.20: 25207393-25207393 on Build GRCh38
rs778142512 Chr.20: 25207327-25207327 on Build GRCh38
rs550929700 Chr.20: 25207244-25207244 on Build GRCh38
rs148914334 Chr.20: 25207258-25207258 on Build GRCh38
rs748500454 Chr.20: 25207245-25207245 on Build GRCh38

View Full Product Details