Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46779990-46780215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESD
Entrez Gene ID
2098
Gene Name
esterase D
Gene Aliases
FGH
UniGene
Hs.432491
Gene Chromosome Location
Chr.13: 46771256-46797681 on Build GRCh38
Transcripts
NM_001984.1 XM_005266278.2 XM_011534954.1 XM_017020414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
432445 c.511G>C p.A171P Chr.13: 46780024-46780024 on Build GRCh38
1661716 c.523A>C p.I175L Chr.13: 46780012-46780012 on Build GRCh38
5695738 c.542G>T p.C181F Chr.13: 46779993-46779993 on Build GRCh38
4850961 c.545C>A p.P182H Chr.13: 46779990-46779990 on Build GRCh38
3987423 c.519T>C p.A173A Chr.13: 46780016-46780016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748131726 Chr.13: 46780023-46780023 on Build GRCh38
rs573859006 Chr.13: 46780183-46780183 on Build GRCh38
rs1039020096 Chr.13: 46780195-46780195 on Build GRCh38
rs779391844 Chr.13: 46780019-46780019 on Build GRCh38
rs752507205 Chr.13: 46780117-46780117 on Build GRCh38
rs912942158 Chr.13: 46780147-46780147 on Build GRCh38
rs763089211 Chr.13: 46780067-46780067 on Build GRCh38
rs765438784 Chr.13: 46780048-46780048 on Build GRCh38
rs773122451 Chr.13: 46780039-46780039 on Build GRCh38
rs771837482 Chr.13: 46780030-46780030 on Build GRCh38
rs775599639 Chr.13: 46780054-46780054 on Build GRCh38
rs945645232 Chr.13: 46780053-46780053 on Build GRCh38
rs760389358 Chr.13: 46780041-46780041 on Build GRCh38
rs765472856 Chr.13: 46780122-46780122 on Build GRCh38
rs374314394 Chr.13: 46780012-46780012 on Build GRCh38

View Full Product Details