Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41159029-41159302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6
Entrez Gene ID
729
Gene Name
complement component 6
Gene Aliases
-
UniGene
Hs.481992
Gene Chromosome Location
Chr.5: 41142116-41261486 on Build GRCh38
Transcripts
NM_000065.3 NM_001115131.2 XM_005248357.2 XM_006714496.3 XM_011514114.2 XM_011514115.2 XM_011514116.2 XM_011514117.2 XM_011514118.2 XM_011514119.2 XM_011514121.2 XM_017009818.1 XM_017009819.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7329722 c.1817G>A p.R606Q Chr.5: 41159121-41159121 on Build GRCh38
73953 c.1726A>C p.T576P Chr.5: 41159212-41159212 on Build GRCh38
6616358 c.1783_1784insC p.Q595fs*8 Chr.5: 41159154-41159155 on Build GRCh38
172891 c.1798C>T p.R600C Chr.5: 41159140-41159140 on Build GRCh38
5639259 c.1823A>T p.E608V Chr.5: 41159115-41159115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770706271 Chr.5: 41159255-41159255 on Build GRCh38
rs200375246 Chr.5: 41159171-41159171 on Build GRCh38
rs752015265 Chr.5: 41159098-41159099 on Build GRCh38
rs76528010 Chr.5: 41159090-41159090 on Build GRCh38
rs760471024 Chr.5: 41159121-41159121 on Build GRCh38
rs776817489 Chr.5: 41159179-41159179 on Build GRCh38
rs1036985798 Chr.5: 41159158-41159158 on Build GRCh38
rs982276270 Chr.5: 41159113-41159113 on Build GRCh38
rs191386155 Chr.5: 41159122-41159122 on Build GRCh38
rs756691542 Chr.5: 41159140-41159140 on Build GRCh38
rs767932073 Chr.5: 41159204-41159204 on Build GRCh38
rs763257632 Chr.5: 41159187-41159187 on Build GRCh38
rs759549524 Chr.5: 41159246-41159246 on Build GRCh38
rs771814711 Chr.5: 41159106-41159106 on Build GRCh38
rs752169165 Chr.5: 41159148-41159148 on Build GRCh38
rs111663292 Chr.5: 41159081-41159081 on Build GRCh38
rs745746350 Chr.5: 41159277-41159277 on Build GRCh38
rs747732468 Chr.5: 41159161-41159161 on Build GRCh38
rs1044498258 Chr.5: 41159129-41159129 on Build GRCh38
rs372194362 Chr.5: 41159242-41159242 on Build GRCh38
rs759873837 Chr.5: 41159139-41159139 on Build GRCh38
rs151080428 Chr.5: 41159153-41159153 on Build GRCh38
rs79523005 Chr.5: 41159243-41159243 on Build GRCh38
rs142881576 Chr.5: 41159152-41159152 on Build GRCh38
rs754102602 Chr.5: 41159067-41159067 on Build GRCh38
rs754488543 Chr.5: 41159233-41159233 on Build GRCh38
rs759707776 Chr.5: 41159184-41159184 on Build GRCh38
rs753075707 Chr.5: 41159077-41159077 on Build GRCh38
rs758842975 Chr.5: 41159145-41159145 on Build GRCh38
rs749038461 Chr.5: 41159091-41159091 on Build GRCh38
rs773204275 Chr.5: 41159114-41159114 on Build GRCh38
rs778566926 Chr.5: 41159160-41159160 on Build GRCh38
rs760051109 Chr.5: 41159136-41159137 on Build GRCh38
rs965256888 Chr.5: 41159162-41159162 on Build GRCh38
rs747031933 Chr.5: 41159174-41159174 on Build GRCh38
rs768267421 Chr.5: 41159093-41159093 on Build GRCh38
rs760839300 Chr.5: 41159055-41159055 on Build GRCh38
rs541865390 Chr.5: 41159201-41159201 on Build GRCh38
rs746754447 Chr.5: 41159250-41159250 on Build GRCh38
rs369711642 Chr.5: 41159186-41159186 on Build GRCh38
rs371329890 Chr.5: 41159224-41159224 on Build GRCh38
rs141061499 Chr.5: 41159191-41159191 on Build GRCh38
rs207465951 Chr.5: 41159178-41159178 on Build GRCh38
rs781283242 Chr.5: 41159276-41159276 on Build GRCh38
rs545271270 Chr.5: 41159156-41159156 on Build GRCh38
rs747412547 Chr.5: 41159084-41159084 on Build GRCh38
rs1032022596 Chr.5: 41159068-41159068 on Build GRCh38
rs764310313 Chr.5: 41159146-41159146 on Build GRCh38
rs755665447 Chr.5: 41159089-41159089 on Build GRCh38
rs770058820 Chr.5: 41159279-41159279 on Build GRCh38
rs764589870 Chr.5: 41159190-41159190 on Build GRCh38
rs148521858 Chr.5: 41159151-41159151 on Build GRCh38
rs776707501 Chr.5: 41159169-41159169 on Build GRCh38
rs957972609 Chr.5: 41159070-41159070 on Build GRCh38
rs778335591 Chr.5: 41159236-41159236 on Build GRCh38
rs776567083 Chr.5: 41159130-41159130 on Build GRCh38
rs80108105 Chr.5: 41159237-41159237 on Build GRCh38
rs765761919 Chr.5: 41159075-41159075 on Build GRCh38
rs905925905 Chr.5: 41159136-41159136 on Build GRCh38
rs753142196 Chr.5: 41159239-41159242 on Build GRCh38
rs753409567 Chr.5: 41159128-41159128 on Build GRCh38
rs747958045 Chr.5: 41159102-41159102 on Build GRCh38
rs112425670 Chr.5: 41159080-41159080 on Build GRCh38
rs767886734 Chr.5: 41159154-41159155 on Build GRCh38
rs766300783 Chr.5: 41159060-41159060 on Build GRCh38
rs1053317922 Chr.5: 41159266-41159266 on Build GRCh38
rs755234921 Chr.5: 41159073-41159073 on Build GRCh38
rs756462651 Chr.5: 41159269-41159272 on Build GRCh38

View Full Product Details