Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231226894-231227119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf131
Entrez Gene ID
128061
Gene Name
chromosome 1 open reading frame 131
Gene Aliases
-
UniGene
Hs.743464
Gene Chromosome Location
Chr.1: 231223763-231241187 on Build GRCh38
Transcripts
NM_001300830.1 NM_152379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6056387 c.583C>T p.R195C Chr.1: 231226996-231226996 on Build GRCh38
5637335 c.580G>A p.E194K Chr.1: 231226999-231226999 on Build GRCh38
4029438 c.543G>A p.T181T Chr.1: 231227036-231227036 on Build GRCh38
7212495 c.511-5C>T p.? Chr.1: 231227073-231227073 on Build GRCh38
1738453 c.604A>G p.K202E Chr.1: 231226975-231226975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777680896 Chr.1: 231226969-231226969 on Build GRCh38
rs770333573 Chr.1: 231227094-231227095 on Build GRCh38
rs751202650 Chr.1: 231227034-231227034 on Build GRCh38
rs866488265 Chr.1: 231226954-231226954 on Build GRCh38
rs374394519 Chr.1: 231226940-231226940 on Build GRCh38
rs751203151 Chr.1: 231226952-231226952 on Build GRCh38
rs372589031 Chr.1: 231226961-231226961 on Build GRCh38
rs750009026 Chr.1: 231226937-231226937 on Build GRCh38
rs144671689 Chr.1: 231227036-231227036 on Build GRCh38
rs775969071 Chr.1: 231226996-231226996 on Build GRCh38
rs147296791 Chr.1: 231227065-231227065 on Build GRCh38
rs754755378 Chr.1: 231226955-231226955 on Build GRCh38
rs762570323 Chr.1: 231227028-231227028 on Build GRCh38
rs757358771 Chr.1: 231227059-231227059 on Build GRCh38
rs201778363 Chr.1: 231227093-231227093 on Build GRCh38
rs199848098 Chr.1: 231227072-231227072 on Build GRCh38
rs756128260 Chr.1: 231226967-231226967 on Build GRCh38
rs781001502 Chr.1: 231226958-231226958 on Build GRCh38
rs373942539 Chr.1: 231227037-231227037 on Build GRCh38
rs762374270 Chr.1: 231227074-231227074 on Build GRCh38
rs34759016 Chr.1: 231227049-231227049 on Build GRCh38
rs764673164 Chr.1: 231226925-231226925 on Build GRCh38
rs779000270 Chr.1: 231227061-231227061 on Build GRCh38
rs199713175 Chr.1: 231227070-231227070 on Build GRCh38
rs749372964 Chr.1: 231226978-231226978 on Build GRCh38
rs200812484 Chr.1: 231227002-231227002 on Build GRCh38
rs528039065 Chr.1: 231227050-231227050 on Build GRCh38
rs765943386 Chr.1: 231227032-231227032 on Build GRCh38
rs140297588 Chr.1: 231227094-231227094 on Build GRCh38
rs1049984453 Chr.1: 231226982-231226982 on Build GRCh38
rs772150625 Chr.1: 231226995-231226995 on Build GRCh38
rs542541652 Chr.1: 231227064-231227064 on Build GRCh38
rs755838517 Chr.1: 231227045-231227045 on Build GRCh38
rs766136833 Chr.1: 231226944-231226944 on Build GRCh38
rs1040612644 Chr.1: 231226965-231226965 on Build GRCh38
rs779129150 Chr.1: 231226985-231226985 on Build GRCh38
rs777214409 Chr.1: 231227010-231227010 on Build GRCh38
rs892025292 Chr.1: 231227038-231227038 on Build GRCh38
rs901714938 Chr.1: 231226977-231226977 on Build GRCh38
rs746058153 Chr.1: 231226993-231226993 on Build GRCh38
rs777304006 Chr.1: 231227073-231227073 on Build GRCh38
rs1020729567 Chr.1: 231226990-231226990 on Build GRCh38
rs139640765 Chr.1: 231226979-231226979 on Build GRCh38

View Full Product Details