Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18101940-18102173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRG2
Entrez Gene ID
1819
Gene Name
developmentally regulated GTP binding protein 2
Gene Aliases
-
UniGene
Hs.78582
Gene Chromosome Location
Chr.17: 18087867-18107985 on Build GRCh38
Transcripts
NM_001388.4 XM_005256499.2 XM_005256500.2 XM_011523704.1 XM_011523705.1 XM_011523706.1 XM_011523707.1 XM_017024296.1 XM_017024297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064442 c.781C>T p.R261* Chr.17: 18101972-18101972 on Build GRCh38
1709991 c.788C>T p.P263L Chr.17: 18101979-18101979 on Build GRCh38
4064441 c.752T>C p.I251T Chr.17: 18101943-18101943 on Build GRCh38
1381273 c.753C>T p.I251I Chr.17: 18101944-18101944 on Build GRCh38
417160 c.763G>A p.E255K Chr.17: 18101954-18101954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746904743 Chr.17: 18101985-18101985 on Build GRCh38
rs998058680 Chr.17: 18102060-18102060 on Build GRCh38
rs1050909628 Chr.17: 18102066-18102066 on Build GRCh38
rs550942772 Chr.17: 18102030-18102030 on Build GRCh38
rs1022711930 Chr.17: 18102128-18102128 on Build GRCh38
rs779652802 Chr.17: 18102103-18102103 on Build GRCh38
rs539577394 Chr.17: 18102123-18102123 on Build GRCh38
rs892820295 Chr.17: 18102090-18102090 on Build GRCh38
rs748006011 Chr.17: 18102013-18102013 on Build GRCh38
rs577964196 Chr.17: 18102129-18102129 on Build GRCh38
rs909221945 Chr.17: 18101963-18101963 on Build GRCh38
rs901906890 Chr.17: 18102037-18102037 on Build GRCh38
rs1045369579 Chr.17: 18102056-18102056 on Build GRCh38
rs189164573 Chr.17: 18101964-18101964 on Build GRCh38
rs948296732 Chr.17: 18102006-18102006 on Build GRCh38
rs1020060273 Chr.17: 18102104-18102104 on Build GRCh38
rs889748457 Chr.17: 18102067-18102067 on Build GRCh38
rs940659477 Chr.17: 18102014-18102014 on Build GRCh38
rs748954433 Chr.17: 18102045-18102045 on Build GRCh38
rs780999702 Chr.17: 18102011-18102011 on Build GRCh38
rs368597699 Chr.17: 18102041-18102041 on Build GRCh38
rs1032897583 Chr.17: 18102089-18102089 on Build GRCh38
rs758537189 Chr.17: 18101973-18101973 on Build GRCh38
rs150706450 Chr.17: 18102124-18102124 on Build GRCh38
rs180690885 Chr.17: 18102086-18102086 on Build GRCh38
rs372057719 Chr.17: 18102044-18102044 on Build GRCh38
rs201062067 Chr.17: 18101983-18101983 on Build GRCh38
rs754894145 Chr.17: 18101990-18101990 on Build GRCh38

View Full Product Details