Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67438341-67438568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1B
Entrez Gene ID
57175
Gene Name
coronin 1B
Gene Aliases
CORONIN-2
UniGene
Hs.6191
Gene Chromosome Location
Chr.11: 67438047-67443821 on Build GRCh38
Transcripts
NM_001018070.2 NM_020441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3355818 c.1432G>C p.E478Q Chr.11: 67438414-67438414 on Build GRCh38
1470562 c.1385C>A p.A462D Chr.11: 67438461-67438461 on Build GRCh38
4525977 c.1366G>A p.V456M Chr.11: 67438480-67438480 on Build GRCh38
2041965 c.1426C>T p.R476C Chr.11: 67438420-67438420 on Build GRCh38
6634223 c.1439A>T p.Q480L Chr.11: 67438407-67438407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200669952 Chr.11: 67438476-67438476 on Build GRCh38
rs376326514 Chr.11: 67438463-67438463 on Build GRCh38
rs377490435 Chr.11: 67438388-67438388 on Build GRCh38
rs370588808 Chr.11: 67438530-67438530 on Build GRCh38
rs747479600 Chr.11: 67438545-67438545 on Build GRCh38
rs777268553 Chr.11: 67438396-67438396 on Build GRCh38
rs1023360609 Chr.11: 67438369-67438369 on Build GRCh38
rs756759787 Chr.11: 67438451-67438451 on Build GRCh38
rs140306387 Chr.11: 67438468-67438468 on Build GRCh38
rs776871409 Chr.11: 67438480-67438480 on Build GRCh38
rs755848771 Chr.11: 67438519-67438519 on Build GRCh38
rs567152204 Chr.11: 67438464-67438464 on Build GRCh38
rs202196580 Chr.11: 67438379-67438379 on Build GRCh38
rs745532568 Chr.11: 67438455-67438455 on Build GRCh38
rs144165811 Chr.11: 67438478-67438478 on Build GRCh38
rs377142759 Chr.11: 67438362-67438362 on Build GRCh38
rs538873958 Chr.11: 67438505-67438505 on Build GRCh38
rs928345885 Chr.11: 67438392-67438392 on Build GRCh38
rs766564864 Chr.11: 67438366-67438366 on Build GRCh38
rs190944233 Chr.11: 67438452-67438452 on Build GRCh38
rs1044754430 Chr.11: 67438525-67438525 on Build GRCh38
rs150750630 Chr.11: 67438484-67438484 on Build GRCh38
rs775434287 Chr.11: 67438471-67438471 on Build GRCh38
rs776587502 Chr.11: 67438539-67438539 on Build GRCh38
rs769763204 Chr.11: 67438407-67438407 on Build GRCh38
rs766589867 Chr.11: 67438420-67438420 on Build GRCh38
rs183838147 Chr.11: 67438499-67438499 on Build GRCh38
rs750575676 Chr.11: 67438361-67438361 on Build GRCh38
rs776215043 Chr.11: 67438399-67438399 on Build GRCh38
rs763019519 Chr.11: 67438412-67438412 on Build GRCh38
rs372523755 Chr.11: 67438409-67438409 on Build GRCh38
rs1056424572 Chr.11: 67438488-67438488 on Build GRCh38
rs775522312 Chr.11: 67438411-67438411 on Build GRCh38
rs201349226 Chr.11: 67438531-67438531 on Build GRCh38
rs2286624 Chr.11: 67438419-67438419 on Build GRCh38
rs554963461 Chr.11: 67438541-67438541 on Build GRCh38
rs376930864 Chr.11: 67438429-67438429 on Build GRCh38
rs770975907 Chr.11: 67438534-67438534 on Build GRCh38
rs147206539 Chr.11: 67438432-67438432 on Build GRCh38
rs754054235 Chr.11: 67438421-67438421 on Build GRCh38
rs755334931 Chr.11: 67438384-67438384 on Build GRCh38
rs143727296 Chr.11: 67438398-67438398 on Build GRCh38
rs745647442 Chr.11: 67438401-67438401 on Build GRCh38
rs570272003 Chr.11: 67438387-67438387 on Build GRCh38
rs763375504 Chr.11: 67438495-67438495 on Build GRCh38
rs750033602 Chr.11: 67438513-67438513 on Build GRCh38
rs746526416 Chr.11: 67438450-67438450 on Build GRCh38
rs769999537 Chr.11: 67438548-67438548 on Build GRCh38
rs377161519 Chr.11: 67438527-67438527 on Build GRCh38
rs201115607 Chr.11: 67438475-67438475 on Build GRCh38
rs778858291 Chr.11: 67438532-67438532 on Build GRCh38
rs530415676 Chr.11: 67438433-67438433 on Build GRCh38
rs3203703 Chr.11: 67438405-67438405 on Build GRCh38
rs755285194 Chr.11: 67438428-67438428 on Build GRCh38
rs759758364 Chr.11: 67438483-67438483 on Build GRCh38
rs375600023 Chr.11: 67438533-67438533 on Build GRCh38
rs767642247 Chr.11: 67438418-67438418 on Build GRCh38
rs752057952 Chr.11: 67438501-67438501 on Build GRCh38
rs749149418 Chr.11: 67438474-67438474 on Build GRCh38
rs753046692 Chr.11: 67438491-67438491 on Build GRCh38
rs872375 Chr.11: 67438507-67438507 on Build GRCh38

View Full Product Details