Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38346427-38346686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERG
Entrez Gene ID
57828
Gene Name
cation channel sperm associated auxiliary subunit gamma
Gene Aliases
C19orf15
UniGene
Hs.324335
Gene Chromosome Location
Chr.19: 38335563-38370949 on Build GRCh38
Transcripts
NM_021185.4 XM_005259114.4 XM_005259115.4 XM_005259116.2 XM_006723308.3 XM_006723310.2 XM_006723311.2 XM_011527174.2 XM_011527175.2 XM_011527176.2 XM_011527177.2 XM_011527178.2 XM_011527179.1 XM_011527182.2 XM_017027053.1 XM_017027054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6539165 c.816T>G p.S272S Chr.19: 38346596-38346596 on Build GRCh38
1612170 c.801+6C>T p.? Chr.19: 38346632-38346632 on Build GRCh38
5936441 c.665C>T p.S222L Chr.19: 38346490-38346490 on Build GRCh38
4644205 c.775G>A p.V259M Chr.19: 38346600-38346600 on Build GRCh38
4599056 c.715G>A p.G239S Chr.19: 38346540-38346540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373768754 Chr.19: 38346544-38346544 on Build GRCh38
rs772623482 Chr.19: 38346611-38346611 on Build GRCh38
rs777833749 Chr.19: 38346514-38346514 on Build GRCh38
rs745526433 Chr.19: 38346550-38346550 on Build GRCh38
rs778874926 Chr.19: 38346467-38346467 on Build GRCh38
rs551833511 Chr.19: 38346507-38346507 on Build GRCh38
rs761177473 Chr.19: 38346633-38346633 on Build GRCh38
rs750164549 Chr.19: 38346458-38346458 on Build GRCh38
rs772711111 Chr.19: 38346653-38346653 on Build GRCh38
rs903845409 Chr.19: 38346500-38346500 on Build GRCh38
rs762140406 Chr.19: 38346600-38346600 on Build GRCh38
rs376729186 Chr.19: 38346599-38346599 on Build GRCh38
rs369750909 Chr.19: 38346446-38346446 on Build GRCh38
rs780691154 Chr.19: 38346526-38346526 on Build GRCh38
rs770347780 Chr.19: 38346523-38346523 on Build GRCh38
rs766382677 Chr.19: 38346638-38346638 on Build GRCh38
rs781467936 Chr.19: 38346561-38346561 on Build GRCh38
rs193036151 Chr.19: 38346459-38346459 on Build GRCh38
rs752503122 Chr.19: 38346469-38346469 on Build GRCh38
rs907126920 Chr.19: 38346491-38346491 on Build GRCh38
rs758453597 Chr.19: 38346513-38346513 on Build GRCh38
rs1031506484 Chr.19: 38346529-38346529 on Build GRCh38
rs531329932 Chr.19: 38346517-38346517 on Build GRCh38
rs773703930 Chr.19: 38346632-38346632 on Build GRCh38
rs763377806 Chr.19: 38346518-38346518 on Build GRCh38
rs966284144 Chr.19: 38346664-38346664 on Build GRCh38
rs769445715 Chr.19: 38346560-38346560 on Build GRCh38
rs755618383 Chr.19: 38346643-38346646 on Build GRCh38
rs955955075 Chr.19: 38346553-38346553 on Build GRCh38
rs764322906 Chr.19: 38346655-38346655 on Build GRCh38

View Full Product Details