Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8125458-8125731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4A
Entrez Gene ID
50856
Gene Name
C-type lectin domain family 4 member A
Gene Aliases
CD367, CLECSF6, DCIR, DDB27, HDCGC13P, LLIR
UniGene
Hs.504657
Gene Chromosome Location
Chr.12: 8102932-8138607 on Build GRCh38
Transcripts
NM_016184.3 NM_194447.2 NM_194448.2 NM_194450.2 XM_017019382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1993786 c.179C>G p.S60* Chr.12: 8125657-8125657 on Build GRCh38
7361511 c.121G>A p.G41R Chr.12: 8125599-8125599 on Build GRCh38
5841965 c.128C>T p.P43L Chr.12: 8125606-8125606 on Build GRCh38
4389535 c.199+7A>T p.? Chr.12: 8125684-8125684 on Build GRCh38
7361512 c.119C>G p.T40S Chr.12: 8125597-8125597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112445640 Chr.12: 8125597-8125597 on Build GRCh38
rs117213717 Chr.12: 8125599-8125599 on Build GRCh38
rs750356261 Chr.12: 8125609-8125609 on Build GRCh38
rs779911427 Chr.12: 8125626-8125626 on Build GRCh38
rs73244076 Chr.12: 8125646-8125646 on Build GRCh38
rs775662970 Chr.12: 8125569-8125569 on Build GRCh38
rs201088323 Chr.12: 8125648-8125648 on Build GRCh38
rs764185718 Chr.12: 8125595-8125595 on Build GRCh38
rs771585431 Chr.12: 8125518-8125518 on Build GRCh38
rs910939711 Chr.12: 8125566-8125566 on Build GRCh38
rs368914199 Chr.12: 8125621-8125621 on Build GRCh38
rs2024301 Chr.12: 8125585-8125585 on Build GRCh38
rs760015006 Chr.12: 8125549-8125549 on Build GRCh38
rs754688932 Chr.12: 8125633-8125633 on Build GRCh38
rs971883539 Chr.12: 8125524-8125524 on Build GRCh38
rs746369204 Chr.12: 8125677-8125677 on Build GRCh38
rs375614072 Chr.12: 8125571-8125571 on Build GRCh38
rs768918126 Chr.12: 8125711-8125711 on Build GRCh38
rs751245632 Chr.12: 8125628-8125628 on Build GRCh38
rs139327535 Chr.12: 8125652-8125652 on Build GRCh38
rs368276318 Chr.12: 8125708-8125708 on Build GRCh38
rs774973868 Chr.12: 8125523-8125523 on Build GRCh38
rs775958662 Chr.12: 8125706-8125706 on Build GRCh38
rs760036177 Chr.12: 8125605-8125605 on Build GRCh38
rs758185932 Chr.12: 8125625-8125625 on Build GRCh38
rs772395608 Chr.12: 8125691-8125691 on Build GRCh38
rs779626131 Chr.12: 8125673-8125673 on Build GRCh38
rs767915121 Chr.12: 8125564-8125564 on Build GRCh38
rs1054556078 Chr.12: 8125581-8125581 on Build GRCh38
rs753939545 Chr.12: 8125598-8125598 on Build GRCh38
rs1049484632 Chr.12: 8125502-8125502 on Build GRCh38

View Full Product Details