Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61013830-61014103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD6
Entrez Gene ID
923
Gene Name
CD6 molecule
Gene Aliases
TP120
UniGene
Hs.744366
Gene Chromosome Location
Chr.11: 60971641-61020377 on Build GRCh38
Transcripts
NM_001254750.1 NM_001254751.1 NM_006725.4 NR_045638.1 XM_006718738.1 XM_006718739.1 XM_006718740.1 XM_006718741.1 XM_011545360.1 XM_011545362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6354656 c.1383A>G p.K461K Chr.11: 61014010-61014010 on Build GRCh38
4980346 c.1362G>T p.P454P Chr.11: 61013989-61013989 on Build GRCh38
7183145 c.1371C>T p.I457I Chr.11: 61013998-61013998 on Build GRCh38
3450635 c.1336C>T p.P446S Chr.11: 61013963-61013963 on Build GRCh38
1604850 c.1380C>A p.P460P Chr.11: 61014007-61014007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756003923 Chr.11: 61014045-61014045 on Build GRCh38
rs747958938 Chr.11: 61013951-61013951 on Build GRCh38
rs764654089 Chr.11: 61013982-61013982 on Build GRCh38
rs762536148 Chr.11: 61013880-61013880 on Build GRCh38
rs764562469 Chr.11: 61013909-61013909 on Build GRCh38
rs549973863 Chr.11: 61013878-61013879 on Build GRCh38
rs752472697 Chr.11: 61013999-61013999 on Build GRCh38
rs892970140 Chr.11: 61013992-61013992 on Build GRCh38
rs754259701 Chr.11: 61013917-61013917 on Build GRCh38
rs567700137 Chr.11: 61013926-61013926 on Build GRCh38
rs549860628 Chr.11: 61013890-61013890 on Build GRCh38
rs751653190 Chr.11: 61014028-61014028 on Build GRCh38
rs184940498 Chr.11: 61013870-61013870 on Build GRCh38
rs752631879 Chr.11: 61014042-61014042 on Build GRCh38
rs1045199214 Chr.11: 61014036-61014036 on Build GRCh38
rs753195993 Chr.11: 61013853-61013853 on Build GRCh38
rs146232748 Chr.11: 61013877-61013877 on Build GRCh38
rs750692888 Chr.11: 61014023-61014023 on Build GRCh38
rs33999602 Chr.11: 61013940-61013941 on Build GRCh38
rs1012656009 Chr.11: 61013993-61013993 on Build GRCh38
rs781268732 Chr.11: 61013950-61013950 on Build GRCh38
rs773119380 Chr.11: 61013952-61013954 on Build GRCh38
rs759074109 Chr.11: 61013981-61013981 on Build GRCh38
rs200544030 Chr.11: 61014021-61014021 on Build GRCh38
rs199919045 Chr.11: 61013886-61013886 on Build GRCh38
rs761126680 Chr.11: 61013901-61013901 on Build GRCh38
rs373752284 Chr.11: 61013891-61013891 on Build GRCh38
rs765540646 Chr.11: 61014008-61014008 on Build GRCh38
rs10897135 Chr.11: 61013918-61013918 on Build GRCh38
rs201598086 Chr.11: 61013965-61013965 on Build GRCh38
rs781285558 Chr.11: 61014035-61014035 on Build GRCh38
rs755045794 Chr.11: 61013911-61013912 on Build GRCh38
rs139338052 Chr.11: 61014069-61014069 on Build GRCh38
rs757614899 Chr.11: 61013924-61013924 on Build GRCh38
rs1038042842 Chr.11: 61013984-61013984 on Build GRCh38
rs146783809 Chr.11: 61013964-61013964 on Build GRCh38
rs1003735379 Chr.11: 61014077-61014077 on Build GRCh38
rs371522495 Chr.11: 61014051-61014051 on Build GRCh38
rs376880399 Chr.11: 61013989-61013989 on Build GRCh38
rs368928420 Chr.11: 61013963-61013963 on Build GRCh38
rs1020086839 Chr.11: 61013890-61013890 on Build GRCh38
rs770585653 Chr.11: 61013971-61013971 on Build GRCh38
rs112948328 Chr.11: 61013928-61013928 on Build GRCh38
rs773782815 Chr.11: 61013974-61013974 on Build GRCh38
rs747085155 Chr.11: 61013945-61013945 on Build GRCh38
rs766606373 Chr.11: 61014025-61014025 on Build GRCh38
rs373128989 Chr.11: 61013988-61013988 on Build GRCh38
rs769652140 Chr.11: 61013961-61013961 on Build GRCh38
rs112922308 Chr.11: 61013919-61013919 on Build GRCh38
rs376920970 Chr.11: 61013927-61013927 on Build GRCh38
rs1049619033 Chr.11: 61014059-61014059 on Build GRCh38
rs928351755 Chr.11: 61013966-61013966 on Build GRCh38
rs758532281 Chr.11: 61013938-61013938 on Build GRCh38
rs148791898 Chr.11: 61013947-61013947 on Build GRCh38

View Full Product Details