Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11003025-11003298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC16A
Entrez Gene ID
23274
Gene Name
C-type lectin domain family 16 member A
Gene Aliases
Gop-1, KIAA0350
UniGene
Hs.35490
Gene Chromosome Location
Chr.16: 10944488-11193272 on Build GRCh38
Transcripts
NM_001243403.1 NM_015226.2 XM_005255210.1 XM_005255211.1 XM_005255213.1 XM_005255214.1 XM_005255215.3 XM_005255216.1 XM_006720870.3 XM_011522434.1 XM_011522435.1 XM_011522436.2 XM_011522437.2 XM_011522438.2 XM_011522439.2 XM_011522440.2 XM_017023089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6143551 c.1169G>C p.R390T Chr.16: 11003171-11003171 on Build GRCh38
3387176 c.1180G>A p.V394I Chr.16: 11003188-11003188 on Build GRCh38
3664769 c.1072-5C>T p.? Chr.16: 11003069-11003069 on Build GRCh38
5850145 c.1073C>T p.P358L Chr.16: 11003081-11003081 on Build GRCh38
4335839 c.1186_1188delGAA p.E398delE Chr.16: 11003194-11003196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007156592 Chr.16: 11003202-11003202 on Build GRCh38
rs957696151 Chr.16: 11003254-11003254 on Build GRCh38
rs745883424 Chr.16: 11003083-11003083 on Build GRCh38
rs74163612 Chr.16: 11003175-11003175 on Build GRCh38
rs748372135 Chr.16: 11003263-11003263 on Build GRCh38
rs375251864 Chr.16: 11003242-11003242 on Build GRCh38
rs1016979068 Chr.16: 11003229-11003229 on Build GRCh38
rs747277078 Chr.16: 11003163-11003163 on Build GRCh38
rs74163613 Chr.16: 11003245-11003245 on Build GRCh38
rs867693563 Chr.16: 11003121-11003121 on Build GRCh38
rs776428030 Chr.16: 11003194-11003196 on Build GRCh38
rs759451816 Chr.16: 11003189-11003189 on Build GRCh38
rs762716460 Chr.16: 11003181-11003181 on Build GRCh38
rs780212753 Chr.16: 11003244-11003244 on Build GRCh38
rs201942249 Chr.16: 11003224-11003224 on Build GRCh38
rs779618011 Chr.16: 11003154-11003154 on Build GRCh38
rs781337434 Chr.16: 11003253-11003253 on Build GRCh38
rs200592669 Chr.16: 11003089-11003089 on Build GRCh38
rs770709204 Chr.16: 11003187-11003187 on Build GRCh38
rs774312800 Chr.16: 11003188-11003188 on Build GRCh38
rs370305113 Chr.16: 11003234-11003234 on Build GRCh38
rs199542538 Chr.16: 11003069-11003069 on Build GRCh38
rs752111516 Chr.16: 11003194-11003194 on Build GRCh38
rs767040613 Chr.16: 11003192-11003192 on Build GRCh38
rs74163611 Chr.16: 11003123-11003123 on Build GRCh38
rs201741967 Chr.16: 11003261-11003261 on Build GRCh38
rs1018442592 Chr.16: 11003131-11003131 on Build GRCh38
rs749551593 Chr.16: 11003139-11003139 on Build GRCh38
rs772830418 Chr.16: 11003108-11003108 on Build GRCh38
rs778234518 Chr.16: 11003138-11003138 on Build GRCh38
rs998279087 Chr.16: 11003171-11003171 on Build GRCh38
rs780883782 Chr.16: 11003059-11003059 on Build GRCh38
rs765126265 Chr.16: 11003097-11003097 on Build GRCh38
rs376930549 Chr.16: 11003098-11003098 on Build GRCh38
rs754552566 Chr.16: 11003118-11003118 on Build GRCh38
rs199821701 Chr.16: 11003227-11003227 on Build GRCh38
rs534658099 Chr.16: 11003148-11003148 on Build GRCh38
rs555591433 Chr.16: 11003091-11003091 on Build GRCh38
rs370740805 Chr.16: 11003075-11003075 on Build GRCh38
rs766249160 Chr.16: 11003112-11003112 on Build GRCh38
rs930064719 Chr.16: 11003119-11003119 on Build GRCh38
rs199858196 Chr.16: 11003193-11003193 on Build GRCh38
rs768769569 Chr.16: 11003251-11003251 on Build GRCh38
rs757760220 Chr.16: 11003232-11003232 on Build GRCh38
rs752447152 Chr.16: 11003122-11003122 on Build GRCh38
rs770762293 Chr.16: 11003265-11003265 on Build GRCh38
rs775920891 Chr.16: 11003090-11003090 on Build GRCh38
rs977864050 Chr.16: 11003205-11003205 on Build GRCh38
rs200410772 Chr.16: 11003153-11003153 on Build GRCh38
rs760382405 Chr.16: 11003196-11003196 on Build GRCh38
rs759504859 Chr.16: 11003270-11003270 on Build GRCh38
rs773061498 Chr.16: 11003177-11003177 on Build GRCh38
rs572081883 Chr.16: 11003230-11003230 on Build GRCh38
rs751015915 Chr.16: 11003241-11003241 on Build GRCh38
rs550022722 Chr.16: 11003054-11003054 on Build GRCh38
rs1006225001 Chr.16: 11003130-11003130 on Build GRCh38
rs765893167 Chr.16: 11003053-11003053 on Build GRCh38
rs771304504 Chr.16: 11003082-11003082 on Build GRCh38
rs201475050 Chr.16: 11003109-11003109 on Build GRCh38
rs200623353 Chr.16: 11003195-11003195 on Build GRCh38
rs747798116 Chr.16: 11003068-11003068 on Build GRCh38
rs775688073 Chr.16: 11003162-11003162 on Build GRCh38
rs565835071 Chr.16: 11003115-11003115 on Build GRCh38
rs201074148 Chr.16: 11003159-11003159 on Build GRCh38
rs374674725 Chr.16: 11003158-11003158 on Build GRCh38

View Full Product Details