Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66938206-66938451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES2
Entrez Gene ID
8824
Gene Name
carboxylesterase 2
Gene Aliases
CE-2, CES2A1, PCE-2, iCE
UniGene
Hs.282975
Gene Chromosome Location
Chr.16: 66934444-66945096 on Build GRCh38
Transcripts
NM_003869.5 NM_198061.2 NR_036684.1 XM_011523421.2 XM_017023818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3957710 c.455G>T p.G152V Chr.16: 66938223-66938223 on Build GRCh38
4061713 c.468G>A p.P156P Chr.16: 66938236-66938236 on Build GRCh38
3510874 c.466C>T p.P156S Chr.16: 66938234-66938234 on Build GRCh38
6988661 c.467C>T p.P156L Chr.16: 66938235-66938235 on Build GRCh38
3818231 c.447G>A p.V149V Chr.16: 66938215-66938215 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376611588 Chr.16: 66938336-66938337 on Build GRCh38
rs769814646 Chr.16: 66938288-66938288 on Build GRCh38
rs571443920 Chr.16: 66938414-66938414 on Build GRCh38
rs529330765 Chr.16: 66938322-66938322 on Build GRCh38
rs958161556 Chr.16: 66938369-66938369 on Build GRCh38
rs757548111 Chr.16: 66938235-66938235 on Build GRCh38
rs548984906 Chr.16: 66938349-66938349 on Build GRCh38
rs748472520 Chr.16: 66938275-66938275 on Build GRCh38
rs746232972 Chr.16: 66938237-66938237 on Build GRCh38
rs768777060 Chr.16: 66938267-66938267 on Build GRCh38
rs944298187 Chr.16: 66938424-66938424 on Build GRCh38
rs780556873 Chr.16: 66938248-66938248 on Build GRCh38
rs759427651 Chr.16: 66938284-66938284 on Build GRCh38
rs769890848 Chr.16: 66938255-66938279 on Build GRCh38
rs911282928 Chr.16: 66938406-66938406 on Build GRCh38
rs751271883 Chr.16: 66938233-66938233 on Build GRCh38
rs772189576 Chr.16: 66938277-66938277 on Build GRCh38
rs539420272 Chr.16: 66938398-66938398 on Build GRCh38
rs1027119780 Chr.16: 66938269-66938269 on Build GRCh38
rs554641992 Chr.16: 66938428-66938428 on Build GRCh38
rs773405319 Chr.16: 66938255-66938255 on Build GRCh38
rs747450152 Chr.16: 66938236-66938236 on Build GRCh38
rs748447942 Chr.16: 66938234-66938234 on Build GRCh38
rs113960944 Chr.16: 66938392-66938392 on Build GRCh38
rs756550589 Chr.16: 66938247-66938247 on Build GRCh38
rs568884325 Chr.16: 66938379-66938379 on Build GRCh38
rs1031126355 Chr.16: 66938367-66938367 on Build GRCh38
rs776431742 Chr.16: 66938282-66938282 on Build GRCh38
rs975727045 Chr.16: 66938429-66938429 on Build GRCh38
rs983008703 Chr.16: 66938316-66938316 on Build GRCh38
rs778992426 Chr.16: 66938273-66938273 on Build GRCh38
rs929855226 Chr.16: 66938242-66938242 on Build GRCh38
rs796150914 Chr.16: 66938332-66938332 on Build GRCh38
rs372933978 Chr.16: 66938263-66938263 on Build GRCh38
rs775608065 Chr.16: 66938290-66938290 on Build GRCh38
rs999733431 Chr.16: 66938366-66938366 on Build GRCh38
rs1014315460 Chr.16: 66938343-66938343 on Build GRCh38

View Full Product Details