Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67874726-67874951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPTF
Entrez Gene ID
2186
Gene Name
bromodomain PHD finger transcription factor
Gene Aliases
FAC1, FALZ, NURF301
UniGene
Hs.444200
Gene Chromosome Location
Chr.17: 67825517-67984378 on Build GRCh38
Transcripts
NM_004459.6 NM_182641.3 XM_005257150.3 XM_005257151.3 XM_005257152.2 XM_005257153.3 XM_005257154.3 XM_005257155.3 XM_005257156.3 XM_005257157.3 XM_005257158.3 XM_005257159.2 XM_005257160.2 XM_005257161.3 XM_011524520.2 XM_011524521.2 XM_011524522.2 XM_011524523.2 XM_011524525.2 XM_011524526.2 XM_017024353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370868 c.1756G>C p.D586H Chr.17: 67874912-67874912 on Build GRCh38
6612426 c.1700T>C p.I567T Chr.17: 67874856-67874856 on Build GRCh38
5522199 c.1672G>A p.E558K Chr.17: 67874828-67874828 on Build GRCh38
127445 c.1746G>A p.E582E Chr.17: 67874902-67874902 on Build GRCh38
3370867 c.1756G>C p.D586H Chr.17: 67874912-67874912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042507503 Chr.17: 67874833-67874833 on Build GRCh38
rs1028025979 Chr.17: 67874908-67874908 on Build GRCh38
rs774586985 Chr.17: 67874909-67874909 on Build GRCh38
rs146878308 Chr.17: 67874784-67874784 on Build GRCh38
rs891908013 Chr.17: 67874795-67874795 on Build GRCh38
rs781047955 Chr.17: 67874856-67874856 on Build GRCh38
rs868522803 Chr.17: 67874894-67874894 on Build GRCh38
rs927495466 Chr.17: 67874764-67874764 on Build GRCh38
rs777795424 Chr.17: 67874834-67874834 on Build GRCh38
rs772739109 Chr.17: 67874888-67874888 on Build GRCh38
rs773906752 Chr.17: 67874781-67874781 on Build GRCh38
rs760372683 Chr.17: 67874860-67874865 on Build GRCh38
rs375245234 Chr.17: 67874817-67874817 on Build GRCh38
rs752572545 Chr.17: 67874794-67874797 on Build GRCh38
rs891998749 Chr.17: 67874808-67874808 on Build GRCh38
rs771413164 Chr.17: 67874903-67874903 on Build GRCh38
rs759962872 Chr.17: 67874914-67874914 on Build GRCh38
rs768565860 Chr.17: 67874855-67874855 on Build GRCh38
rs750355395 Chr.17: 67874836-67874836 on Build GRCh38
rs1010423532 Chr.17: 67874818-67874818 on Build GRCh38
rs367775856 Chr.17: 67874853-67874853 on Build GRCh38
rs779723761 Chr.17: 67874848-67874848 on Build GRCh38
rs769469510 Chr.17: 67874883-67874883 on Build GRCh38
rs751904000 Chr.17: 67874804-67874804 on Build GRCh38
rs747778763 Chr.17: 67874870-67874870 on Build GRCh38
rs769414401 Chr.17: 67874772-67874772 on Build GRCh38
rs182255249 Chr.17: 67874805-67874805 on Build GRCh38
rs767362095 Chr.17: 67874789-67874790 on Build GRCh38
rs371490474 Chr.17: 67874789-67874789 on Build GRCh38
rs756247991 Chr.17: 67874831-67874831 on Build GRCh38
rs758305005 Chr.17: 67874843-67874843 on Build GRCh38
rs763394557 Chr.17: 67874898-67874898 on Build GRCh38
rs752915826 Chr.17: 67874823-67874823 on Build GRCh38

View Full Product Details