Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17766634-17766901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
UniGene
Hs.96485
Gene Chromosome Location
Chr.19: 17747718-17788568 on Build GRCh38
Transcripts
NM_001161357.1 NM_001161358.1 NM_001161359.1 NM_015122.2 XM_005259831.3 XM_006722701.3 XM_006722702.3 XM_011527824.2 XM_011527825.2 XM_011527826.2 XM_011527827.2 XM_011527828.2 XM_017026519.1 XM_017026520.1 XM_017026521.1 XM_017026522.1 XM_017026523.1 XM_017026524.1 XM_017026525.1 XM_017026526.1 XM_017026527.1 XM_017026528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7112224 c.211_216delTGGGAG p.W71_E72delWE Chr.19: 17766685-17766690 on Build GRCh38
4476446 c.206C>T p.P69L Chr.19: 17766680-17766680 on Build GRCh38
281115 c.228C>A p.V76V Chr.19: 17766702-17766702 on Build GRCh38
1206937 c.208C>T p.L70F Chr.19: 17766682-17766682 on Build GRCh38
4140389 c.264G>C p.L88L Chr.19: 17766738-17766738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774043293 Chr.19: 17766752-17766752 on Build GRCh38
rs966279633 Chr.19: 17766746-17766746 on Build GRCh38
rs761605670 Chr.19: 17766653-17766653 on Build GRCh38
rs985922422 Chr.19: 17766676-17766676 on Build GRCh38
rs138314971 Chr.19: 17766765-17766765 on Build GRCh38
rs372337585 Chr.19: 17766826-17766826 on Build GRCh38
rs368695542 Chr.19: 17766822-17766822 on Build GRCh38
rs745675840 Chr.19: 17766720-17766720 on Build GRCh38
rs535465534 Chr.19: 17766875-17766875 on Build GRCh38
rs779018736 Chr.19: 17766690-17766690 on Build GRCh38
rs370442783 Chr.19: 17766683-17766683 on Build GRCh38
rs767300572 Chr.19: 17766768-17766768 on Build GRCh38
rs750849014 Chr.19: 17766661-17766661 on Build GRCh38
rs954160855 Chr.19: 17766675-17766675 on Build GRCh38
rs765922249 Chr.19: 17766859-17766859 on Build GRCh38
rs756451278 Chr.19: 17766669-17766669 on Build GRCh38
rs767245618 Chr.19: 17766656-17766656 on Build GRCh38
rs763015274 Chr.19: 17766742-17766742 on Build GRCh38
rs749004797 Chr.19: 17766816-17766816 on Build GRCh38
rs896366737 Chr.19: 17766863-17766863 on Build GRCh38
rs771056954 Chr.19: 17766801-17766801 on Build GRCh38
rs199533162 Chr.19: 17766844-17766844 on Build GRCh38
rs772589046 Chr.19: 17766846-17766846 on Build GRCh38
rs772323726 Chr.19: 17766710-17766710 on Build GRCh38
rs767788386 Chr.19: 17766784-17766784 on Build GRCh38
rs376703961 Chr.19: 17766659-17766659 on Build GRCh38
rs750578179 Chr.19: 17766851-17766851 on Build GRCh38
rs755561445 Chr.19: 17766775-17766775 on Build GRCh38
rs768702939 Chr.19: 17766743-17766743 on Build GRCh38
rs374582760 Chr.19: 17766780-17766780 on Build GRCh38
rs777936504 Chr.19: 17766717-17766717 on Build GRCh38
rs142806172 Chr.19: 17766772-17766772 on Build GRCh38
rs754135952 Chr.19: 17766773-17766773 on Build GRCh38
rs775294778 Chr.19: 17766738-17766738 on Build GRCh38
rs202137888 Chr.19: 17766705-17766705 on Build GRCh38
rs572940628 Chr.19: 17766865-17766865 on Build GRCh38
rs759856600 Chr.19: 17766860-17766860 on Build GRCh38
rs146048151 Chr.19: 17766781-17766781 on Build GRCh38
rs760431404 Chr.19: 17766858-17766858 on Build GRCh38
rs769574803 Chr.19: 17766723-17766723 on Build GRCh38
rs750035566 Chr.19: 17766769-17766769 on Build GRCh38
rs200710206 Chr.19: 17766671-17766671 on Build GRCh38
rs779364469 Chr.19: 17766776-17766776 on Build GRCh38
rs780435067 Chr.19: 17766681-17766681 on Build GRCh38
rs564195039 Chr.19: 17766857-17766857 on Build GRCh38
rs1056821293 Chr.19: 17766827-17766827 on Build GRCh38
rs774381291 Chr.19: 17766823-17766823 on Build GRCh38
rs747188478 Chr.19: 17766783-17766783 on Build GRCh38
rs376359634 Chr.19: 17766842-17766842 on Build GRCh38

View Full Product Details