Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 867028-867253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAK
Entrez Gene ID
2580
Gene Name
cyclin G associated kinase
Gene Aliases
DNAJ26, DNAJC26
UniGene
Hs.369607
Gene Chromosome Location
Chr.4: 849275-932390 on Build GRCh38
Transcripts
NM_001318134.1 NM_005255.3 XM_005272268.2 XM_005272270.2 XM_011513425.2 XM_011513426.2 XM_011513427.2 XM_011513428.2 XM_011513429.2 XM_011513430.1 XM_011513431.2 XM_011513432.2 XM_011513434.2 XM_017007991.1 XM_017007992.1 XM_017007993.1 XM_017007994.1 XM_017007995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2956511 c.2674G>A p.A892T Chr.4: 867154-867154 on Build GRCh38
4904409 c.2743C>T p.L915F Chr.4: 867085-867085 on Build GRCh38
6668834 c.2605G>A p.A869T Chr.4: 867223-867223 on Build GRCh38
1431447 c.2673C>T p.G891G Chr.4: 867155-867155 on Build GRCh38
4126473 c.2698G>C p.A900P Chr.4: 867130-867130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201968587 Chr.4: 867070-867070 on Build GRCh38
rs758153369 Chr.4: 867178-867178 on Build GRCh38
rs753898629 Chr.4: 867155-867155 on Build GRCh38
rs542390112 Chr.4: 867067-867067 on Build GRCh38
rs760768090 Chr.4: 867089-867089 on Build GRCh38
rs750175813 Chr.4: 867116-867116 on Build GRCh38
rs776443423 Chr.4: 867055-867055 on Build GRCh38
rs761515694 Chr.4: 867056-867056 on Build GRCh38
rs878886319 Chr.4: 867132-867132 on Build GRCh38
rs550901346 Chr.4: 867190-867190 on Build GRCh38
rs528569570 Chr.4: 867098-867098 on Build GRCh38
rs372718341 Chr.4: 867153-867153 on Build GRCh38
rs149842313 Chr.4: 867198-867198 on Build GRCh38
rs547097269 Chr.4: 867134-867134 on Build GRCh38
rs376010644 Chr.4: 867164-867164 on Build GRCh38
rs1141285 Chr.4: 867210-867210 on Build GRCh38
rs767880649 Chr.4: 867078-867078 on Build GRCh38
rs1022043352 Chr.4: 867062-867062 on Build GRCh38
rs768468619 Chr.4: 867054-867054 on Build GRCh38
rs747607099 Chr.4: 867191-867191 on Build GRCh38
rs757359967 Chr.4: 867104-867104 on Build GRCh38
rs201584204 Chr.4: 867226-867226 on Build GRCh38
rs150706546 Chr.4: 867163-867163 on Build GRCh38
rs779957711 Chr.4: 867180-867180 on Build GRCh38
rs746483074 Chr.4: 867053-867053 on Build GRCh38
rs571457670 Chr.4: 867139-867139 on Build GRCh38
rs769143099 Chr.4: 867225-867225 on Build GRCh38
rs201137296 Chr.4: 867135-867135 on Build GRCh38
rs748930143 Chr.4: 867206-867206 on Build GRCh38
rs113722219 Chr.4: 867224-867224 on Build GRCh38
rs764169845 Chr.4: 867154-867154 on Build GRCh38
rs758405633 Chr.4: 867120-867120 on Build GRCh38
rs368428903 Chr.4: 867147-867147 on Build GRCh38
rs745833929 Chr.4: 867051-867051 on Build GRCh38
rs764120874 Chr.4: 867101-867101 on Build GRCh38
rs754906971 Chr.4: 867185-867185 on Build GRCh38
rs1048558345 Chr.4: 867158-867158 on Build GRCh38
rs771329576 Chr.4: 867058-867058 on Build GRCh38
rs774521502 Chr.4: 867061-867061 on Build GRCh38
rs373562468 Chr.4: 867202-867202 on Build GRCh38
rs866186796 Chr.4: 867050-867050 on Build GRCh38
rs774846126 Chr.4: 867138-867138 on Build GRCh38
rs778823149 Chr.4: 867110-867110 on Build GRCh38
rs892171872 Chr.4: 867222-867222 on Build GRCh38
rs535853224 Chr.4: 867223-867223 on Build GRCh38
rs769317923 Chr.4: 867137-867137 on Build GRCh38
rs760147192 Chr.4: 867068-867068 on Build GRCh38
rs761780920 Chr.4: 867228-867228 on Build GRCh38
rs532200203 Chr.4: 867172-867172 on Build GRCh38
rs752823840 Chr.4: 867084-867084 on Build GRCh38
rs200903151 Chr.4: 867103-867103 on Build GRCh38
rs750285508 Chr.4: 867175-867175 on Build GRCh38
rs139051032 Chr.4: 867184-867184 on Build GRCh38
rs772326344 Chr.4: 867207-867207 on Build GRCh38
rs772431440 Chr.4: 867145-867145 on Build GRCh38

View Full Product Details