Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31657899-31658164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT6
Entrez Gene ID
9331
Gene Name
beta-1,4-galactosyltransferase 6
Gene Aliases
B4Gal-T6, beta4Gal-T6
UniGene
Hs.591063
Gene Chromosome Location
Chr.18: 31622246-31724670 on Build GRCh38
Transcripts
NM_004775.3 XM_005258387.4 XM_006722578.3 XM_006722579.3 XM_017026088.1 XM_017026089.1 XM_017026090.1 XM_017026091.1 XM_017026092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3525265 c.318C>T p.L106L Chr.18: 31658004-31658004 on Build GRCh38
4916114 c.317T>C p.L106P Chr.18: 31658005-31658005 on Build GRCh38
1740442 c.281C>T p.T94M Chr.18: 31658041-31658041 on Build GRCh38
5749052 c.268G>T p.V90F Chr.18: 31658054-31658054 on Build GRCh38
1711171 c.238C>T p.P80S Chr.18: 31658084-31658084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749726741 Chr.18: 31658102-31658102 on Build GRCh38
rs745814574 Chr.18: 31658119-31658120 on Build GRCh38
rs1051441392 Chr.18: 31658131-31658131 on Build GRCh38
rs796552917 Chr.18: 31658112-31658113 on Build GRCh38
rs763875463 Chr.18: 31658074-31658074 on Build GRCh38
rs780680005 Chr.18: 31658098-31658098 on Build GRCh38
rs757053005 Chr.18: 31658115-31658116 on Build GRCh38
rs774420565 Chr.18: 31658029-31658029 on Build GRCh38
rs756724410 Chr.18: 31658092-31658092 on Build GRCh38
rs775641080 Chr.18: 31658102-31658102 on Build GRCh38
rs753655835 Chr.18: 31657941-31657941 on Build GRCh38
rs752343903 Chr.18: 31658114-31658115 on Build GRCh38
rs574354534 Chr.18: 31658041-31658041 on Build GRCh38
rs746319501 Chr.18: 31657984-31657984 on Build GRCh38
rs903338182 Chr.18: 31658036-31658036 on Build GRCh38
rs762742243 Chr.18: 31658130-31658130 on Build GRCh38
rs150412311 Chr.18: 31658032-31658032 on Build GRCh38
rs749635114 Chr.18: 31658019-31658019 on Build GRCh38
rs768894782 Chr.18: 31658107-31658107 on Build GRCh38
rs367718116 Chr.18: 31658040-31658040 on Build GRCh38
rs745798949 Chr.18: 31658094-31658094 on Build GRCh38
rs562316833 Chr.18: 31658001-31658001 on Build GRCh38
rs760721382 Chr.18: 31658122-31658122 on Build GRCh38
rs773208111 Chr.18: 31658120-31658120 on Build GRCh38
rs775404081 Chr.18: 31658129-31658129 on Build GRCh38
rs766393776 Chr.18: 31657931-31657931 on Build GRCh38
rs756819238 Chr.18: 31658138-31658138 on Build GRCh38
rs752381929 Chr.18: 31657952-31657952 on Build GRCh38
rs752537607 Chr.18: 31658055-31658055 on Build GRCh38
rs397704070 Chr.18: 31658127-31658127 on Build GRCh38
rs781154999 Chr.18: 31658116-31658117 on Build GRCh38
rs67738912 Chr.18: 31658114-31658114 on Build GRCh38
rs746407564 Chr.18: 31658085-31658085 on Build GRCh38
rs763737080 Chr.18: 31658135-31658135 on Build GRCh38
rs371001508 Chr.18: 31657933-31657933 on Build GRCh38
rs756803714 Chr.18: 31658081-31658081 on Build GRCh38
rs770343819 Chr.18: 31657969-31657969 on Build GRCh38
rs763174255 Chr.18: 31658107-31658107 on Build GRCh38
rs769805613 Chr.18: 31658124-31658125 on Build GRCh38
rs544511294 Chr.18: 31657968-31657968 on Build GRCh38
rs373869100 Chr.18: 31658031-31658031 on Build GRCh38
rs781724599 Chr.18: 31657981-31657981 on Build GRCh38
rs758178797 Chr.18: 31658063-31658063 on Build GRCh38
rs76171871 Chr.18: 31658082-31658082 on Build GRCh38
rs577025669 Chr.18: 31657965-31657965 on Build GRCh38
rs751245040 Chr.18: 31658137-31658137 on Build GRCh38
rs772228932 Chr.18: 31658113-31658113 on Build GRCh38
rs78832366 Chr.18: 31658116-31658116 on Build GRCh38
rs769044404 Chr.18: 31658027-31658027 on Build GRCh38
rs558336486 Chr.18: 31657942-31657942 on Build GRCh38
rs770303733 Chr.18: 31658003-31658003 on Build GRCh38
rs758088806 Chr.18: 31657961-31657961 on Build GRCh38
rs765129509 Chr.18: 31658050-31658050 on Build GRCh38
rs764374296 Chr.18: 31658112-31658113 on Build GRCh38
rs758132300 Chr.18: 31658113-31658114 on Build GRCh38
rs777637061 Chr.18: 31658114-31658116 on Build GRCh38
rs759639072 Chr.18: 31657926-31657930 on Build GRCh38
rs776490893 Chr.18: 31657926-31657927 on Build GRCh38
rs983757056 Chr.18: 31658042-31658042 on Build GRCh38
rs371863169 Chr.18: 31658111-31658112 on Build GRCh38
rs774649977 Chr.18: 31658109-31658109 on Build GRCh38
rs141770767 Chr.18: 31657992-31657992 on Build GRCh38
rs541497223 Chr.18: 31658080-31658080 on Build GRCh38
rs765217351 Chr.18: 31658128-31658128 on Build GRCh38
rs1032388438 Chr.18: 31657932-31657932 on Build GRCh38
rs914225321 Chr.18: 31657976-31657976 on Build GRCh38
rs921680655 Chr.18: 31658083-31658083 on Build GRCh38
rs769794853 Chr.18: 31657934-31657938 on Build GRCh38
rs372369000 Chr.18: 31658113-31658113 on Build GRCh38
rs748271370 Chr.18: 31658112-31658112 on Build GRCh38

View Full Product Details