Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50351223-50351448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM186A
Entrez Gene ID
121006
Gene Name
family with sequence similarity 186 member A
Gene Aliases
-
UniGene
Hs.133187
Gene Chromosome Location
Chr.12: 50326854-50396633 on Build GRCh38
Transcripts
NM_001145475.1 XM_006719231.3 XM_011537890.2 XM_011537892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5544952 c.5591C>T p.P1864L Chr.12: 50351241-50351241 on Build GRCh38
7416358 c.5443G>A p.A1815T Chr.12: 50351389-50351389 on Build GRCh38
1362244 c.5483C>A p.S1828Y Chr.12: 50351349-50351349 on Build GRCh38
1362243 c.5566_5567delCT p.L1856fs*9 Chr.12: 50351265-50351266 on Build GRCh38
4638162 c.5561C>A p.A1854D Chr.12: 50351271-50351271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559409210 Chr.12: 50351247-50351247 on Build GRCh38
rs927095514 Chr.12: 50351274-50351274 on Build GRCh38
rs936998965 Chr.12: 50351279-50351279 on Build GRCh38
rs367586420 Chr.12: 50351346-50351346 on Build GRCh38
rs544787297 Chr.12: 50351302-50351302 on Build GRCh38
rs747149791 Chr.12: 50351369-50351369 on Build GRCh38
rs867695610 Chr.12: 50351265-50351265 on Build GRCh38
rs890938129 Chr.12: 50351377-50351377 on Build GRCh38
rs1032736573 Chr.12: 50351383-50351383 on Build GRCh38
rs79391678 Chr.12: 50351311-50351311 on Build GRCh38
rs780908996 Chr.12: 50351271-50351271 on Build GRCh38
rs370839385 Chr.12: 50351391-50351391 on Build GRCh38
rs1010171328 Chr.12: 50351357-50351357 on Build GRCh38
rs563668989 Chr.12: 50351429-50351429 on Build GRCh38
rs1041609494 Chr.12: 50351347-50351347 on Build GRCh38
rs1019439169 Chr.12: 50351396-50351396 on Build GRCh38
rs572828458 Chr.12: 50351259-50351259 on Build GRCh38
rs765273273 Chr.12: 50351400-50351400 on Build GRCh38
rs895582215 Chr.12: 50351329-50351329 on Build GRCh38
rs753216652 Chr.12: 50351409-50351409 on Build GRCh38
rs371821511 Chr.12: 50351359-50351359 on Build GRCh38
rs774143221 Chr.12: 50351268-50351268 on Build GRCh38
rs757088591 Chr.12: 50351263-50351263 on Build GRCh38
rs550007629 Chr.12: 50351420-50351420 on Build GRCh38
rs937062799 Chr.12: 50351301-50351301 on Build GRCh38
rs972837102 Chr.12: 50351402-50351402 on Build GRCh38
rs954054474 Chr.12: 50351390-50351390 on Build GRCh38
rs996095712 Chr.12: 50351401-50351401 on Build GRCh38

View Full Product Details